Microscope Cover Glasses
Supplier: NEST SCIENTIFIC
Jump to:
À propos de cet article
Microscope cover glasses are manufactured in 100,000 grade cleaning room with USP 6 grade raw materials.
- Resistance to organic solvent
- Autoclavable
- Can be used in confocal experiment
- High quality glass, non-cytotoxic
- Thickness: 0.16 to 0.19 mm(±0.02 mm)
5 Options Available Below
Product Details & Documents
Microscope cover glasses are manufactured in 100,000 grade cleaning room with USP 6 grade raw materials.
- Resistance to organic solvent
- Autoclavable
- Can be used in confocal experiment
- High quality glass, non-cytotoxic
- Thickness: 0.16 to 0.19 mm(±0.02 mm)
Product Family Options
Informations sur le produit
- PackedDésignationØMatièreAvailabilityPrice
- 100/pack24 well cell culture plate14 mmGlassCase of 100$120.70Each$120.70Supplier Number: 801010
- 100/pack24 well cell culture plate15 mmGlassCase of 100$120.70Each$120.70Supplier Number: 801007
- 100/pack12 well cell culture plate18 mmGlassCase of 100$120.70Each$120.70Supplier Number: 801011
- 100/pack12 well cell culture plate20 mmGlassCase of 100$120.70Each$120.70Supplier Number: 801008
- 100/pack6 well cell culture plate25 mmGlassCase of 100$120.70Each$120.70Supplier Number: 801009