Colormark™ and Colormark™ Plus Microscope Slides, Epredia™
Supplier: Erie Scientific
Certificates
About this item
Slides are designed for the SHUR/Mark® etching labeling system and other automated labeling systems
A contrasting color is printed on the back of the painted end so that the etched patient or specimen information is clearly visible on the front. These slides offer excellent etched labeling visibility. Colormark® Plus slides have undergone an adhesive treatment that holds tissue sections without the need for coating steps in the laboratory. This virtually eliminates tissue loss during staining procedures.
Frequently Bought Together