Cryo Boxes, PC
Supplier: NEST SCIENTIFIC
Jump to:
À propos de cet article
Cryo boxes are manufactured in 100,000 grade cleaning room with USP 6 grade raw materials.
- Temperature range: −196 to +70 ℃
- Individual package
- Clear polycarbonate cover for easy identification
- Alphanumeric grid for easy sample identification
- Each box includes drain holes and air vents to minimize condensation
10 Options Available Below
Product Details & Documents
Cryo boxes are manufactured in 100,000 grade cleaning room with USP 6 grade raw materials.
- Temperature range: −196 to +70 ℃
- Individual package
- Clear polycarbonate cover for easy identification
- Alphanumeric grid for easy sample identification
- Each box includes drain holes and air vents to minimize condensation
Product Family Options
Informations sur le produit
- ColorFor VialsMatièreAvailabilityPrice
- Blue1.9 ml vials, 8×6 arrayPCCase of 10$118.78Each$118.78Supplier Number: 616931
- Blue4.0 ml vials, 8×6 arrayPCCase of 10$129.96Each$129.96Supplier Number: 616932
- Blue0.5 ml vials, 9×9 arrayPCCase of 56$709.02Each$709.02Supplier Number: 616224
- Blue1.2 ml vials, 9×9 arrayPCCase of 48$623.92Each$623.92Supplier Number: 616325
- Blue2.0 ml vials, 9×9 arrayPCCase of 40$629.53Each$629.53Supplier Number: 616021
- Blue3.0 ml vials, 9×9 arrayPCCase of 32$531.76Each$531.76Supplier Number: 616421
- Blue4.0 ml vials, 9×9 arrayPCCase of 32$531.76Each$531.76Supplier Number: 616121
- Blue5.0 ml vials, 9×9 arrayPCCase of 32$567.19Each$567.19Supplier Number: 616521
- Blue0.5 ml vials, internal thread cryogenic vial only, 10×10 arrayPCCase of 56$709.02Each$709.02Supplier Number: 616204
- Blue1.2 ml vials, internal thread cryogenic vial only, 10×10 arrayPCCase of 48$638.05Each$638.05Supplier Number: 616305
- Blue2.0 ml vials, internal thread cryogenic vial only, 10×10 arrayPCDiscontinued-Supplier Number: 616001