Cell Scrapers
Cell Scrapers
Cell Scrapers
Supplier:  NEST SCIENTIFIC
Certificates
2 Options Available Below
Product Details & Documents
Specially designed cell scrapers can be used to scrape and collect cells easily and effectively. Push the handle with slight pressure to change the blade angle. Then push down the handle towards the bottom of the container. Rotate the handle slightly to make the blade twist to right direction.
  • Free rotating blade, makes the blade twist to the right direction
  • Two sizes are available
  • Easy-tear sterile packing
  • Sterilized by E-beam, SAL=10 to 6
  • Non pyrogenic, DNase/RNase free
  • Individually packaged
Documents
Product Family Options
Product Information
  • Handle Length
    Blade Width
    Availability
    Price
  • 220 mm
    13 mm
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 100
    Case of 100 - $144.48
    $144.48
    Log in for availability
    Each
    Each - $144.48
    $144.48
    Case of 100
       $144.48
    Each
       $144.48
    Supplier Number: 710001
  • 280 mm
    20 mm
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 100
    Case of 100 - $162.85
    $162.85
    Log in for availability
    Each
    Each - $162.85
    $162.85
    Case of 100
       $162.85
    Each
       $162.85
    Supplier Number: 710011