Combs for Deep Well Plates
Supplier: NEST SCIENTIFIC
Jump to:
À propos de cet article
5 Options Available Below
Product Details & Documents
Product Family Options
Informations sur le produit
- DésignationAvailabilityPrice
- Combs for magnetic applications, 96 tip, non sterile, 2/pack, 50/caseCase of 50$451.27Each$451.27Supplier Number: 503311
- Combs for magnetic applications, 96 tip, sterile, 2/pack, 50/caseCase of 50$480.19Each$480.19Supplier Number: 503361
- Plastic comb, 8-strip, non sterile, compatible with 503711 and 503761, 2/pack, 50/box, 500/caseCase of 500$665.29Each$665.29Supplier Number: 509211
- Plastic comb, 8-strip, sterile, compatible with 503711 and 503761, 2/pack, 50/box, 500/caseCase of 500$712.19Each$712.19Supplier Number: 509261
- Plastic comb, 24 tip, sterile, compatible with Thermo KingFisher System, 1/pack, 25/caseCase of 25$255.15Each$255.15Supplier Number: 509001