Corning® Plain Micro Slides, Corning | Avantor
Corning® Plain Micro Slides, Corning
Supplier:   Corning
Corning® Plain Micro Slides, Corning
Supplier:  Corning
Certificates
Corning® Plain Micro Slides, Corning
Corning® Plain Micro Slides, Corning
3 Options Available Below
Product Details & Documents
For use in nonsterile cytology and pathology applications
Made of water-white glass for maximum clarity. Precleaned and ready for use. Inspected for chips and rough edges before packaging.
Ordering information: Packaged 1/2 gross per box. Slides are packed vertically for easy lift out.
Product Family Options
Informations sur le produit
  • Désignation
    Épaisseur
    L×l
    Availability
    Price
  • Plain slides
    0.9 - 1.1 mm
    75×25 mm (3×1")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 10
    Case of 10 - $748.00
    $748.00
    Case of 10
       $748.00
    Supplier Number: 2947-75X25
  • Plain slides
    0.9 - 1.0 mm
    75×38 mm (3×1½")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 5
    Case of 5 - $747.00
    $747.00
    Case of 5
       $747.00
    Supplier Number: 2947-75X38
  • Plain slides
    0.9 - 1.0 mm
    75×50 mm (3×2")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 720
    Case of 720 - $779.00
    $779.00
    Prix promotionnel pour les nouveaux clients
    Case of 720
       $779.00
    Supplier Number: 2947-75X50