Pierce™ In-Solution Tryptic Digestion and Guanidination Kit, Thermo Scientific
CAPI89895
:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Description:In-Solution Tryptic Digestion and Guanidation Kit
- Cat. no.:CAPI89895
- No. of Reactions:90
- Supplier No.:89895
Pierce™ In-Solution Tryptic Digestion and Guanidination Kit contains proteomics grade trypsin, buffers and reagents to alkylate and digest proteins and then guanidinate the peptide fragments for mass spectrometry.
- Optimized trypsin digestion producing complete digests of protein ranging from 0.025 to 10 µg
- Optimized guanidination where reaction produces little or no N-terminal modification
- Protein can be digested and guanidinated all in a day
Accurate identification of proteins and analysis of post-translational modifications by mass spectrometry require accurate and complete protein digestion and peptide modification.