VWR® Micro Cover Glasses, Rectangular
VWR® Micro Cover Glasses, Rectangular
VWR® Micro Cover Glasses, Rectangular
VWR® Micro Cover Glasses, Rectangular
These micro cover glasses are made from the best quality glass, and each set of numbered slides is uniform in size and thickness.
  • Quality glass construction
  • Uniform sizes
  • Two-slide thickness options available
The No. 1 slides are 0.13 to 0.17 mm in thickness. The No. 1¹/₂ slides are 0.16 to 0.19 mm in thickness.
 : Slides are packed 1 oz. per box.
Product Family Options
No. 1 Slides
  • Désignation
    L×l
    Numbering
  • Cover glasses, rectangular
    30×22 mm
    #1
    Ounce
    Ounce - $64.18
    $64.18
    Case of 10
    Case of 10 - $641.80
    $641.80
    Ounce
       $64.18
    Case of 10
       $641.80
  • Cover glasses, rectangular
    40×22 mm
    #1
  • Cover glasses, rectangular
    50×22 mm
    #1
    Ounce
    Ounce - $47.95
    $47.95
    Case of 10
    Case of 10 - $479.50
    $479.50
    Ounce
       $47.95
    Case of 10
       $479.50
  • Cover glasses, rectangular
    50×24 mm
    #1
    Ounce
    Ounce - $42.49
    $42.49
    Case of 10
    Case of 10 - $424.90
    $424.90
    Ounce
       $42.49
    Case of 10
       $424.90
  • Cover glasses, rectangular
    30×24 mm
    #1
    Ounce
    Ounce - $41.69
    $41.69
    Case of 10
    Case of 10 - $416.90
    $416.90
    Ounce
       $41.69
    Case of 10
       $416.90
  • Cover glasses, rectangular
    60×24 mm
    #1
    Ounce
    Ounce - $42.61
    $42.61
    Case of 10
    Case of 10 - $426.10
    $426.10
    Ounce
       $42.61
    Case of 10
       $426.10
  • Cover glasses, rectangular
    40×24 mm
    #1
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $49.34
    $49.34
    Ounce
    Ounce - $49.88
    $49.88
    Case of 10 (1/ Itm/Bx)
    Case of 10 (1/ Itm/Bx) - $498.80
    $498.80
    Each(1/Itm/Bx)
       $49.34
    Ounce
       $49.88
    Case of 10 (1/ Itm/Bx)
       $498.80
No. 1 1/2 Slides
  • Désignation
    L×l
    Numbering
  • Cover glasses, rectangular
    40×22 mm
    #1.5
    Ounce
    Ounce - $42.63
    $42.63
    Case of 10
    Case of 10 - $426.30
    $426.30
    Ounce
       $42.63
    Case of 10
       $426.30
  • Cover glasses, rectangular
    50×24 mm
    #1.5
    Ounce
    Ounce - $41.38
    $41.38
    Case of 10 (1oz)
    Case of 10 (1oz) - $413.80
    $413.80
    Ounce
       $41.38
    Case of 10 (1oz)
       $413.80
  • Cover glasses, rectangular
    60×24 mm
    #1.5
  • Micro cover glasses, rectangular
    30×22 mm
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $41.01
    $41.01
    Ounce
    Ounce - $41.46
    $41.46
    Case of 10 (1/ Itm/Bx)
    Case of 10 (1/ Itm/Bx) - $414.56
    $414.56
    Each(1/Itm/Bx)
       $41.01
    Ounce
       $41.46
    Case of 10 (1/ Itm/Bx)
       $414.56
  • Micro cover glasses, rectangular
    50×22 mm
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $49.94
    $49.94
    Ounce
    Ounce - $50.50
    $50.50
    Case of 10 (1/ Itm/Bx)
    Case of 10 (1/ Itm/Bx) - $504.96
    $504.96
    Each(1/Itm/Bx)
       $49.94
    Ounce
       $50.50
    Case of 10 (1/ Itm/Bx)
       $504.96
  • Micro cover glasses, rectangular
    60×22 mm
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $46.62
    $46.62
    Ounce
    Ounce - $47.13
    $47.13
    Case of 10 (1/ Itm/Bx)
    Case of 10 (1/ Itm/Bx) - $471.28
    $471.28
    Each(1/Itm/Bx)
       $46.62
    Ounce
       $47.13
    Case of 10 (1/ Itm/Bx)
       $471.28
  • Micro cover glasses, rectangular
    30×24 mm
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $50.78
    $50.78
    Ounce
    Ounce - $51.34
    $51.34
    Case of 10 (1/ Itm/Bx)
    Case of 10 (1/ Itm/Bx) - $513.44
    $513.44
    Each(1/Itm/Bx)
       $50.78
    Ounce
       $51.34
    Case of 10 (1/ Itm/Bx)
       $513.44
  • Cover glasses, rectangular
    40×24 mm
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $48.17
    $48.17
    Ounce
    Ounce - $48.70
    $48.70
    Case of 10 (1/ Itm/Bx)
    Case of 10 (1/ Itm/Bx) - $486.96
    $486.96
    Each(1/Itm/Bx)
       $48.17
    Ounce
       $48.70
    Case of 10 (1/ Itm/Bx)
       $486.96