VWR® Solid Rubber Stoppers, Black | Avantor
VWR® Solid Rubber Stoppers, Black
VWR® Solid Rubber Stoppers, Black
Supplier:  VWR International
Certificates
VWR® Solid Rubber Stoppers, Black
VWR® Solid Rubber Stoppers, Black
26 Options Available Below
Product Details & Documents
These solid, black stoppers contain a high percentage of pure rubber, weigh less than neoprene, and are low in free sulfur (1/2%).
  • Resilient and pliable for a long time
  • Low in free sulfur
  • Can withstand adverse laboratory conditions without hardening
Composition ensures resiliency and pliability over a long period.
Packaging: Packaging Information: 26 stoppers per pack.
Product Family Options
Product Information
  • Taille
    Bottom Diameter
    Top Diameter
    Longueur
    Packaging
    Availability
    Price
  • 000
    8 mm
    13 mm
    21 mm
    181/pack
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 1
    Pack of 1 - $272.26
    $272.26
    New Customer Promotional Price
    Log in for availability
    Case of 5
    Case of 5 - $1,361.32
    $1,361.32
    New Customer Promotional Price
    Pack of 1
       $272.26
    Case of 5
       $1,361.32
  • 00
    10 mm
    15 mm
    25 mm
    103/pack
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 1
    Pack of 1 - $187.74
    $187.74
    New Customer Promotional Price
    Log in for availability
    Case of 5
    Case of 5 - $938.72
    $938.72
    New Customer Promotional Price
    Pack of 1
       $187.74
    Case of 5
       $938.72
  • 0
    13 mm
    17 mm
    25 mm
    76/pack
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 1
    Pack of 1 - $168.32
    $168.32
    New Customer Promotional Price
    Log in for availability
    Case of 5
    Case of 5 - $841.60
    $841.60
    New Customer Promotional Price
    Pack of 1
       $168.32
    Case of 5
       $841.60
  • 1
    14 mm
    19 mm
    25 mm
    64/pack
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 1
    Pack of 1 - $178.88
    $178.88
    New Customer Promotional Price
    Log in for availability
    Case of 5
    Case of 5 - $894.40
    $894.40
    New Customer Promotional Price
    Pack of 1
       $178.88
    Case of 5
       $894.40
  • 2
    16 mm
    20 mm
    25 mm
    53/pack
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 1
    Pack of 1 - $182.35
    $182.35
    New Customer Promotional Price
    Log in for availability
    Case of 5
    Case of 5 - $911.76
    $911.76
    New Customer Promotional Price
    Pack of 1
       $182.35
    Case of 5
       $911.76
  • 3
    18 mm
    24 mm
    25 mm
    40/pack
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 1
    Pack of 1 - $218.26
    $218.26
    New Customer Promotional Price
    Log in for availability
    Case of 5
    Case of 5 - $1,091.28
    $1,091.28
    New Customer Promotional Price
    Pack of 1
       $218.26
    Case of 5
       $1,091.28
  • 4
    20 mm
    26 mm
    25 mm
    34/pack
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 35
    Pack of 35 - $226.63
    $226.63
    New Customer Promotional Price
    Log in for availability
    Case of 175
    Case of 175 - $1,133.16
    $1,133.16
    New Customer Promotional Price
    Pack of 35
       $226.63
    Case of 175
       $1,133.16
  • 5
    23 mm
    27 mm
    25 mm
    29/pack
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 1
    Pack of 1 - $238.38
    $238.38
    New Customer Promotional Price
    Log in for availability
    Case of 5
    Case of 5 - $1,191.88
    $1,191.88
    New Customer Promotional Price
    Pack of 1
       $238.38
    Case of 5
       $1,191.88
  • 51/2
    24 mm
    28 mm
    25 mm
    26/pack
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 1
    Pack of 1 - $231.35
    $231.35
    New Customer Promotional Price
    Log in for availability
    Case of 5
    Case of 5 - $1,156.76
    $1,156.76
    New Customer Promotional Price
    Pack of 1
       $231.35
    Case of 5
       $1,156.76
  • 6
    26 mm
    32 mm
    25 mm
    22/pack
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 1
    Pack of 1 - $235.90
    $235.90
    New Customer Promotional Price
    Log in for availability
    Case of 5
    Case of 5 - $1,179.48
    $1,179.48
    New Customer Promotional Price
    Pack of 1
       $235.90
    Case of 5
       $1,179.48
  • 61/2
    27 mm
    34 mm
    25 mm
    20/pack
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 1
    Pack of 1 - $254.86
    $254.86
    New Customer Promotional Price
    Log in for availability
    Case of 5
    Case of 5 - $1,274.28
    $1,274.28
    New Customer Promotional Price
    Pack of 1
       $254.86
    Case of 5
       $1,274.28
  • 7
    30 mm
    37 mm
    25 mm
    16/pack
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 1
    Pack of 1 - $222.65
    $222.65
    New Customer Promotional Price
    Log in for availability
    Case of 5
    Case of 5 - $1,113.24
    $1,113.24
    New Customer Promotional Price
    Pack of 1
       $222.65
    Case of 5
       $1,113.24
  • 71/2
    31 mm
    39 mm
    25 mm
    15/pack
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 1
    Pack of 1 - $251.91
    $251.91
    New Customer Promotional Price
    Log in for availability
    Case of 5
    Case of 5 - $1,259.56
    $1,259.56
    New Customer Promotional Price
    Pack of 1
       $251.91
    Case of 5
       $1,259.56
  • 8
    33 mm
    41 mm
    25 mm
    13/pack
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 1
    Pack of 1 - $223.71
    $223.71
    New Customer Promotional Price
    Log in for availability
    Case of 5 (1lb)
    Case of 5 (1lb) - $1,118.56
    $1,118.56
    New Customer Promotional Price
    Pack of 1
       $223.71
    Case of 5 (1lb)
       $1,118.56
  • 81/2
    36 mm
    43 mm
    25 mm
    12/pack
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 12
    Pack of 12 - $202.06
    $202.06
    New Customer Promotional Price
    Log in for availability
    Case of 60
    Case of 60 - $1,010.32
    $1,010.32
    New Customer Promotional Price
    Pack of 12
       $202.06
    Case of 60
       $1,010.32
  • 9
    37 mm
    45 mm
    25 mm
    11/pack
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 1 (1/Itm/Bx)
    Pack of 1 (1/Itm/Bx) - $213.78
    $213.78
    New Customer Promotional Price
    Log in for availability
    Case of 5 (1/ Itm/Bx)
    Case of 5 (1/ Itm/Bx) - $1,068.88
    $1,068.88
    New Customer Promotional Price
    Pack of 1 (1/Itm/Bx)
       $213.78
    Case of 5 (1/ Itm/Bx)
       $1,068.88
  • 91/2
    38 mm
    46 mm
    25 mm
    10/pack
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 10
    Pack of 10 - $206.82
    $206.82
    New Customer Promotional Price
    Log in for availability
    Case of 50
    Case of 50 - $1,034.12
    $1,034.12
    New Customer Promotional Price
    Pack of 10
       $206.82
    Case of 50
       $1,034.12
  • 10
    42 mm
    50 mm
    25 mm
    9/pack
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 1
    Pack of 1 - $241.78
    $241.78
    New Customer Promotional Price
    Log in for availability
    Case of 5
    Case of 5 - $1,208.88
    $1,208.88
    New Customer Promotional Price
    Pack of 1
       $241.78
    Case of 5
       $1,208.88
  • 101/2
    45 mm
    53 mm
    25 mm
    8/pack
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 1
    Pack of 1 - $218.51
    $218.51
    New Customer Promotional Price
    Log in for availability
    Case of 5
    Case of 5 - $1,092.56
    $1,092.56
    New Customer Promotional Price
    Pack of 1
       $218.51
    Case of 5
       $1,092.56
  • 11
    48 mm
    56 mm
    25 mm
    7/pack
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 1
    Pack of 1 - $240.11
    $240.11
    New Customer Promotional Price
    Log in for availability
    Case of 5
    Case of 5 - $1,200.56
    $1,200.56
    New Customer Promotional Price
    Pack of 1
       $240.11
    Case of 5
       $1,200.56
  • 111/2
    50 mm
    63 mm
    25 mm
    6/pack
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 1
    Pack of 1 - $273.11
    $273.11
    New Customer Promotional Price
    Log in for availability
    Case of 5
    Case of 5 - $1,365.56
    $1,365.56
    New Customer Promotional Price
    Pack of 1
       $273.11
    Case of 5
       $1,365.56
  • 12
    54 mm
    64 mm
    25 mm
    6/pack
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 1
    Pack of 1 - $256.41
    $256.41
    New Customer Promotional Price
    Log in for availability
    Case of 5
    Case of 5 - $1,282.04
    $1,282.04
    New Customer Promotional Price
    Pack of 1
       $256.41
    Case of 5
       $1,282.04
  • 13
    58 mm
    68 mm
    25 mm
    5/pack
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 1
    Pack of 1 - $243.66
    $243.66
    New Customer Promotional Price
    Log in for availability
    Case of 5
    Case of 5 - $1,218.32
    $1,218.32
    New Customer Promotional Price
    Pack of 1
       $243.66
    Case of 5
       $1,218.32
  • 131/2
    62 mm
    75 mm
    35 mm
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $112.57
    $112.57
    New Customer Promotional Price
    Log in for availability
    Case of 13
    Case of 13 - $1,463.43
    $1,463.43
    New Customer Promotional Price
    Each
       $112.57
    Case of 13
       $1,463.43
  • 14
    75 mm
    90 mm
    39 mm
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $126.91
    $126.91
    New Customer Promotional Price
    Log in for availability
    Case of 8
    Case of 8 - $1,015.30
    $1,015.30
    New Customer Promotional Price
    Each
       $126.91
    Case of 8
       $1,015.30
  • 15
    83 mm
    103 mm
    39 mm
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $196.68
    $196.68
    New Customer Promotional Price
    Log in for availability
    Case of 8
    Case of 8 - $1,573.44
    $1,573.44
    New Customer Promotional Price
    Each
       $196.68
    Case of 8
       $1,573.44