VWR® Corks
VWR® Corks
   Sustainable Options Available
Supplier:   VWR International
VWR® Corks
Supplier:  VWR International
   Sustainable Options Available
Certificates
VWR® Corks
28 Options Available Below
Product Details & Documents
XXXX Corks function as a closure for bottles, vials, laboratory and other openings.
  • Regular length taper
  • Assorted sizes
The XXXX premium grade corks have fewer lenticels (crevices) than standard grade
Selected grade of fine textured corks. Sizes 22 through 30 have cemented construction.
This means they are better suited for use as laboratory corks and for transporting many kinds of liquids.
Specifications for Product Family
Specifications
  • Catalog No:
  • 23420-118
  • 23420-162
  • 23420-184
  • 23420-300
  • CA23420-023
  • CA23420-038
  • CA23420-060
  • CA23420-071
  • CA23420-082
  • CA23420-107
  • CA23420-130
  • CA23420-140
  • CA23420-151
  • CA23420-195
  • CA23420-210
  • CA23420-231
  • CA23420-300
  • CA23420-388
  • CA23420-424
  • CA23420-468
  • CA23420-570
  • CA23420-050
  • CA23420-220
  • CA23420-242
  • CA23420-344
  • CA23420-366
  • CA23420-399
  • CA23420-413
  • CA23420-435
  • CA23420-457
  • CA23420-479
  • CA23420-480
  • CA23420-490
  • CA23420-515
  • CA23420-537
  • CA23420-548
  • CA23420-559
  • CA23420-560
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
Specifications
Product Family Options
Product Information
  • Hauteur
    Stopper No.
    Bottom Diameter
    Top Diameter
    Availability
    Price
  • 17 mm
    2
    9 mm
    13 mm
    Discontinued
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    23420-118
    Environmentally Preferable
    Low Manufacturing Impact
      Low Manufacturing Impact
  • 22 mm
    5
    13 mm
    17 mm
    Discontinued
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    23420-162
    Environmentally Preferable
    Low Manufacturing Impact
      Low Manufacturing Impact
  • 24 mm
    6
    14 mm
    19 mm
    Discontinued
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    23420-184
    Environmentally Preferable
    Low Manufacturing Impact
      Low Manufacturing Impact
  • 31 mm
    12
    23 mm
    28 mm
    Discontinued
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    23420-300
    Environmentally Preferable
    Low Manufacturing Impact
      Low Manufacturing Impact
  • 13 mm
    0000
    2 mm
    5 mm
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $65.42
    $65.42
    Pack of 100
       $65.42
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    CA23420-023
    Environmentally Preferable
    Low Manufacturing Impact
      Low Manufacturing Impact
  • 12 mm
    000
    4 mm
    6 mm
    Log in for availability
    Log in for availability
    Pack of 500
    Pack of 500 - $312.71
    $312.71
    Pack of 500
       $312.71
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    CA23420-038
    Environmentally Preferable
    Low Manufacturing Impact
      Low Manufacturing Impact
  • 13 mm
    0
    7 mm
    10 mm
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $79.82
    $79.82
    Pack of 100
       $79.82
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    CA23420-060
    Environmentally Preferable
    Low Manufacturing Impact
      Low Manufacturing Impact
  • 13 mm
    0
    7 mm
    10 mm
    Log in for availability
    Log in for availability
    Pack of 500
    Pack of 500 - $312.71
    $312.71
    Pack of 500
       $312.71
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    CA23420-071
    Environmentally Preferable
    Low Manufacturing Impact
      Low Manufacturing Impact
  • 16 mm
    1
    8 mm
    11 mm
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $79.82
    $79.82
    Pack of 100
       $79.82
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    CA23420-082
    Environmentally Preferable
    Low Manufacturing Impact
      Low Manufacturing Impact
  • 17 mm
    2
    9 mm
    13 mm
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $95.49
    $95.49
    Pack of 100
       $95.49
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    CA23420-107
    Environmentally Preferable
    Low Manufacturing Impact
      Low Manufacturing Impact
  • 18 mm
    3
    11 mm
    14 mm
    Log in for availability
    Log in for availability
    Pack of 500
    Pack of 500 - $389.28
    $389.28
    Pack of 500
       $389.28
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    CA23420-130
    Environmentally Preferable
    Low Manufacturing Impact
      Low Manufacturing Impact
  • 20 mm
    4
    12 mm
    16 mm
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $100.83
    $100.83
    Pack of 100
       $100.83
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    CA23420-140
    Environmentally Preferable
    Low Manufacturing Impact
      Low Manufacturing Impact
  • 20 mm
    4
    12 mm
    16 mm
    Log in for availability
    Log in for availability
    Pack of 500
    Pack of 500 - $395.92
    $395.92
    Pack of 500
       $395.92
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    CA23420-151
    Environmentally Preferable
    Low Manufacturing Impact
      Low Manufacturing Impact
  • 24 mm
    6
    14 mm
    19 mm
    Log in for availability
    Log in for availability
    Pack of 500
    Pack of 500 - $479.48
    $479.48
    Pack of 500
       $479.48
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    CA23420-195
    Environmentally Preferable
    Low Manufacturing Impact
      Low Manufacturing Impact
  • 25 mm
    7
    16 mm
    21 mm
    Log in for availability
    Log in for availability
    Pack of 500
    Pack of 500 - $499.93
    $499.93
    Pack of 500
       $499.93
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    CA23420-210
    Environmentally Preferable
    Low Manufacturing Impact
      Low Manufacturing Impact
  • 26 mm
    8
    17 mm
    22 mm
    Log in for availability
    Log in for availability
    Pack of 500
    Pack of 500 - $950.51
    $950.51
    Pack of 500
       $950.51
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    CA23420-231
    Environmentally Preferable
    Low Manufacturing Impact
      Low Manufacturing Impact
  • 31 mm
    12
    23 mm
    28 mm
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $522.83
    $522.83
    Pack of 100
       $522.83
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    CA23420-300
    Environmentally Preferable
    Low Manufacturing Impact
      Low Manufacturing Impact
  • 38 mm
    16
    28 mm
    35 mm
    Log in for availability
    Log in for availability
    Pack of 10
    Pack of 10 - $94.84
    $94.84
    Pack of 10
       $94.84
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    CA23420-388
    Environmentally Preferable
    Low Manufacturing Impact
      Low Manufacturing Impact
  • 38 mm
    18
    30 mm
    38 mm
    Log in for availability
    Log in for availability
    Pack of 10
    Pack of 10 - $133.99
    $133.99
    Pack of 10
       $133.99
    Specifications:
    More
    Specifications
    Cat. No.
    CA23420-424
    Environmentally Preferable
  • 38 mm
    20
    33 mm
    41 mm
    Log in for availability
    Log in for availability
    Pack of 10
    Pack of 10 - $177.50
    $177.50
    Pack of 10
       $177.50
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    CA23420-468
    Environmentally Preferable
    Low Manufacturing Impact
      Low Manufacturing Impact
  • 38 mm
    30
    50 mm
    57 mm
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $3,450.72
    $3,450.72
    Pack of 100
       $3,450.72
    Specifications:
    More
    Specifications
    Cat. No.
    CA23420-570
    Environmentally Preferable
  • 13 mm
    00
    6 mm
    8 mm
    Log in for availability
    Log in for availability
    Pack of 500
    Pack of 500 - $364.83
    $364.83
    Pack of 500
       $364.83
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    CA23420-050
    Environmentally Preferable
    Low Manufacturing Impact
      Low Manufacturing Impact
  • 26 mm
    8
    17 mm
    22 mm
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $170.99
    $170.99
    Pack of 100
       $170.99
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    CA23420-220
    Environmentally Preferable
    Low Manufacturing Impact
      Low Manufacturing Impact
  • 28 mm
    9
    18 mm
    24 mm
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $209.00
    $209.00
    Pack of 100
       $209.00
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    CA23420-242
    Environmentally Preferable
    Low Manufacturing Impact
      Low Manufacturing Impact
  • 31 mm
    14
    26 mm
    32 mm
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $502.27
    $502.27
    Pack of 100
       $502.27
    Specifications:
    More
    Specifications
    Cat. No.
    CA23420-344
    Environmentally Preferable
  • 31 mm
    15
    27 mm
    33 mm
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $800.00
    $800.00
    Pack of 100
       $800.00
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    CA23420-366
    Environmentally Preferable
    Low Manufacturing Impact
      Low Manufacturing Impact
  • 38 mm
    16
    28 mm
    35 mm
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $864.34
    $864.34
    Pack of 100
       $864.34
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    CA23420-399
    Environmentally Preferable
    Low Manufacturing Impact
      Low Manufacturing Impact
  • 38 mm
    17
    29 mm
    36 mm
    Discontinued
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    CA23420-413
    Environmentally Preferable
    Low Manufacturing Impact
      Low Manufacturing Impact
  • 38 mm
    18
    30 mm
    38 mm
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $1,260.83
    $1,260.83
    Pack of 100
       $1,260.83
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    CA23420-435
    Environmentally Preferable
    Low Manufacturing Impact
      Low Manufacturing Impact
  • 38 mm
    19
    33 mm
    40 mm
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $1,460.33
    $1,460.33
    Pack of 100
       $1,460.33
    Specifications:
    More
    Specifications
    Cat. No.
    CA23420-457
    Environmentally Preferable
  • 38 mm
    20
    33 mm
    41 mm
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $1,449.73
    $1,449.73
    Pack of 100
       $1,449.73
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    CA23420-479
    Environmentally Preferable
    Low Manufacturing Impact
      Low Manufacturing Impact
  • 38 mm
    22
    38 mm
    44 mm
    Discontinued
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    CA23420-480
    Environmentally Preferable
    Low Manufacturing Impact
      Low Manufacturing Impact
  • 38 mm
    22
    38 mm
    44 mm
    Discontinued
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    CA23420-490
    Environmentally Preferable
    Low Manufacturing Impact
      Low Manufacturing Impact
  • 38 mm
    24
    40 mm
    47 mm
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $2,322.99
    $2,322.99
    Pack of 100
       $2,322.99
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    CA23420-515
    Environmentally Preferable
    Low Manufacturing Impact
      Low Manufacturing Impact
  • 38 mm
    26
    44 mm
    50 mm
    Discontinued
    Specifications:
    More
    Specifications
    Cat. No.
    CA23420-537
    Environmentally Preferable
  • 38 mm
    28
    47 mm
    54 mm
    Discontinued
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    CA23420-548
    Environmentally Preferable
    Low Manufacturing Impact
      Low Manufacturing Impact
  • 38 mm
    28
    47 mm
    54 mm
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $3,082.04
    $3,082.04
    Pack of 100
       $3,082.04
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    CA23420-559
    Environmentally Preferable
    Low Manufacturing Impact
      Low Manufacturing Impact
  • Discontinued
    Specifications:
    More
    Specifications
    Cat. No.
    CA23420-560
    Environmentally Preferable