Sample Vials, with HDPE Caps
À propos de cet article
Polypropylene vials, with attached or separated HDPE Caps.
- Self-standing: the vial placed on the lab workbench steadily even without a base
- Sterilized by E-beam, Sterility Assurance Level: SAL 10⁻⁶
- Safe, fast, environment-friendly, no chemical residues
- DNAse-, RNAse-, pyrogen-,endotoxin-free
- Disposable
Excellent sealing performance, no leakage of negative pressure 95 Kpa, ensuring safe and effective air transportation.
Temperature range: Approx.–196 to 121 °C
Certifications: Meets USP VI standard.
13 Options Available Below
Product Details & Documents
Polypropylene vials, with attached or separated HDPE Caps.
- Self-standing: the vial placed on the lab workbench steadily even without a base
- Sterilized by E-beam, Sterility Assurance Level: SAL 10⁻⁶
- Safe, fast, environment-friendly, no chemical residues
- DNAse-, RNAse-, pyrogen-,endotoxin-free
- Disposable
Excellent sealing performance, no leakage of negative pressure 95 Kpa, ensuring safe and effective air transportation.
Temperature range: Approx.–196 to 121 °C
Certifications: Meets USP VI standard.
Product Family Options
With attached caps
Informations sur le produit
- DésignationCapacitéAvec bouchonCap TypeSterilityPackagingAvailabilityPrice
- Sample vial3 mlOuiNormal capSterile50/pack; 1000/caseCase of 1,000$692.45Each$692.45Supplier Number: 624201
- Sample vial4 mlOuiNormal capSterile50/pack; 1000/caseCase of 1,000$692.45Each$692.45Supplier Number: 608201
- Sample vial5 mlOuiNormal capSterile50/pack; 1000/caseCase of 1,000$692.45Each$692.45Supplier Number: 619011
- Sample vial5 mlOuiDouble-threaded screw capSterile50/pack; 1000/caseCase of 1,000$692.45Each$692.45Supplier Number: 619101
- Sample vial5 mlOuiPink capture capSterile50/pack; 1000/caseCase of 1,000$692.45Each$692.45Supplier Number: 619201
- Sample vial10 mlOuiNormal capSterile50/pack; 1000/caseCase of 1,000$800.04Each$800.04Supplier Number: 625001
- Shorter sample vial10 mlOuiNormal capSterile50/pack; 1000/caseCase of 1,000$800.04Each$800.04Supplier Number: 625011
With separated caps
Informations sur le produit
- DésignationCapacitéAvec bouchonCap TypeSterilityPackagingAvailabilityPrice
- Sample vial3 mlOuiNormal capSterile200 tubes/bag, 200 caps/bagCase of 1,000$658.86Each$658.86Supplier Number: 624207
- Sample vial4 mlOuiNormal capSterile200 tubes/bag, 200 caps/bagCase of 1,000$658.86Each$658.86Supplier Number: 608207
- Sample vial5 mlOuiConcave/convex capSterile200 tubes/bag, 200 caps/bagCase of 1,000$658.86Each$658.86Supplier Number: 619017
- Sample vial5 mlOuiDouble-threaded screw capSterile200 tubes/bag, 200 caps/bagCase of 1,000$658.86Each$658.86Supplier Number: 619107
- Sample vial10 mlOuiNormal capSterile200 tubes/bag, 200 caps/bagCase of 1,000$759.67Each$759.67Supplier Number: 625007
- Shorter sample vial10 mlOuiNormal capSterile200 tubes/bag, 200 caps/bagCase of 1,000$759.67Each$759.67Supplier Number: 625017