High-Efficient Erlenmeyer Flasks With Tube
Supplier:   NEST SCIENTIFIC
High-Efficient Erlenmeyer Flasks With Tube
Supplier:  NEST SCIENTIFIC
Certificates
High-Efficient Erlenmeyer Flasks With Tube
13 Options Available Below
Product Details & Documents
The high-efficient Erlenmeyer flask (with tube) is designed to eliminate pipeline design, assembly, sterilization and other operations and improving production efficiency. It is ideal for shaker culture applications and can be used for scale-up culture of suspension cells.
  • Polycarbonate
  • HDPE cap
  • Bi-directional transfer cap with TPE tube
  • Handle: Nylon
  • Sterilized by E-beam, Sterility assurance level: SAL= 10⁻⁶
  • Non-pyrogenic and DNase/RNase free
  • Pipe diameter: ¹/₈" Int.Ø, ¹/₄" Ext.Ø
High-efficiency, large-volume culture flasks allow cells to show strong viability with large expression amount of proteins in the culture of mammalian cells and insect cells. During the culturing process, the use rate of the shaker is significantly increased, and the survival rate and viability of cells are both dramatically.
It is pre-installed on the High efficiency Erlenmeyer flask, which realizes the closed transfer of liquid and reduces the risk of external contamination during the process of capping and screwing. The transfer cover is seal cap and connected with a 33 mm diameter syringe filter with 0.22 μm PTFE.
The transfer cap comes with a large filter membrane, high oxygen flux, and no external air filter is required to ensure high-density culture of cells, while the overall height is reduced, the applicable range of the shaker is wider, and the utilization rate of the shaker is increased.
Product Family Options
Informations sur le produit
  • Désignation
    Capacité
    Sterility
    Longueur Tubes
    Availability
    Price
  • High-efficient Erlenmeyer flask with baffles, bi-directional transfer cap with TPE tube
    2 L
    Sterile
    50 cm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 4
    Case of 4 - $878.69
    $878.69
    Log in pour la disponibilité
    Each
    Each - $878.69
    $878.69
    Case of 4
       $878.69
    Each
       $878.69
    Supplier Number: C20122-AZB050A
  • High-efficient Erlenmeyer flask with baffles, bi-directional transfer cap with TPE tube
    3 L
    Sterile
    50 cm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 4
    Case of 4 - $1,126.60
    $1,126.60
    Log in pour la disponibilité
    Each
    Each - $1,126.60
    $1,126.60
    Case of 4
       $1,126.60
    Each
       $1,126.60
    Supplier Number: C20222-AZB050A
  • High-efficient Erlenmeyer flask with baffles, bi-directional transfer cap with TPE tube, wide-mouth
    3 L
    Sterile
    50 cm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 4
    Case of 4 - $1,126.60
    $1,126.60
    Log in pour la disponibilité
    Each
    Each - $1,126.60
    $1,126.60
    Case of 4
       $1,126.60
    Each
       $1,126.60
    Supplier Number: C20322-AZB050A
  • High-efficient Erlenmeyer flask, bi-directional transfer cap with TPE tube, wide-mouth
    3 L
    Sterile
    50 cm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 4
    Case of 4 - $768.51
    $768.51
    Log in pour la disponibilité
    Each
    Each - $768.51
    $768.51
    Case of 4
       $768.51
    Each
       $768.51
    Supplier Number: C10322-AZB050B
  • High-efficient Erlenmeyer flask, bi-directional transfer cap with TPE tube, wide-mouth
    3 L
    Sterile
    50 cm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 4
    Case of 4 - $1,278.16
    $1,278.16
    Log in pour la disponibilité
    Each
    Each - $1,278.16
    $1,278.16
    Case of 4
       $1,278.16
    Each
       $1,278.16
    Supplier Number: C10323-BZC050A
  • High-efficient Erlenmeyer flask, bi-directional transfer cap with TPE tube
    2 L
    Sterile
    50 cm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 4
    Case of 4 - $1,542.55
    $1,542.55
    Log in pour la disponibilité
    Each
    Each - $1,542.55
    $1,542.55
    Case of 4
       $1,542.55
    Each
       $1,542.55
    Supplier Number: C10122-AZB050A
  • High-efficient Erlenmeyer flask, bi-directional transfer cap with TPE tube
    2 L
    Sterile
    50 cm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 4
    Case of 4 - $1,057.79
    $1,057.79
    Log in pour la disponibilité
    Each
    Each - $1,057.79
    $1,057.79
    Case of 4
       $1,057.79
    Each
       $1,057.79
    Supplier Number: C10123-BZC050A
  • High-efficient Erlenmeyer flask, bi-directional transfer cap with TPE tube
    3 L
    Sterile
    50 cm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 4
    Case of 4 - $1,071.51
    $1,071.51
    Log in pour la disponibilité
    Each
    Each - $1,071.51
    $1,071.51
    Case of 4
       $1,071.51
    Each
       $1,071.51
    Supplier Number: C10222-AZB050A
  • High-efficient Erlenmeyer flask, bi-directional transfer cap with TPE tube
    3 L
    Sterile
    50 cm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 4
    Case of 4 - $1,278.16
    $1,278.16
    Log in pour la disponibilité
    Each
    Each - $1,278.16
    $1,278.16
    Case of 4
       $1,278.16
    Each
       $1,278.16
    Supplier Number: C10223-BZC050A
  • High-efficient Erlenmeyer flask, bi-directional transfer cap with TPE tube
    3 L
    Sterile
    50 cm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 4
    Case of 4 - $1,071.51
    $1,071.51
    Log in pour la disponibilité
    Each
    Each - $1,071.51
    $1,071.51
    Case of 4
       $1,071.51
    Each
       $1,071.51
    Supplier Number: C10322-AZB050A
  • High-efficient Erlenmeyer flask, bi-directional transfer cap with TPE tube
    5 L
    Sterile
    90 cm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 4
    Case of 4 - $1,443.43
    $1,443.43
    Log in pour la disponibilité
    Each
    Each - $1,443.43
    $1,443.43
    Case of 4
       $1,443.43
    Each
       $1,443.43
    Supplier Number: C10422-AZB090A
  • High-efficient Erlenmeyer flask, bi-directional transfer cap with TPE tube
    5 L
    Sterile
    90 cm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 4
    Case of 4 - $1,725.76
    $1,725.76
    Log in pour la disponibilité
    Each
    Each - $1,725.76
    $1,725.76
    Case of 4
       $1,725.76
    Each
       $1,725.76
    Supplier Number: C10423-BZC090A
  • High-efficient Erlenmeyer flask, multifunction transfer cap with TPE tube
    5 L
    Sterile
    30 cm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 4
    Case of 4 - $1,457.15
    $1,457.15
    Log in pour la disponibilité
    Each
    Each - $1,457.15
    $1,457.15
    Case of 4
       $1,457.15
    Each
       $1,457.15
    Supplier Number: C10442-AKA060A