Deep Well Plates
Deep Well Plates
Deep Well Plates
Deep Well Plates
The deep well plates have a variety of specifications, conforming to the needs of fully automatic workstations and experiments.
  • High quality imported PP material employed for high stability and no chemical reactions with test reagents
  • Maximum sustainable centrifuge force 4000×g
  • Compatible with DMSO and inert to water
  • Can be stored under subzero temperature from −40 to −80 ℃
  • Autoclavable at 20 psi, 121 ℃ for 20 minutes, great heating uniformity
  • Minimum residual liquid, low heavy metal content
Genomic DNA extraction, plasmid DNA extraction, and nucleic acid extraction and purification of various samples. High-throughput automatic liquid operation to achieve high-throughput operations, such as protein precipitation, liquid extraction, and processing of animal tissues, bacteria, plants, soil, clinical samples, yeasts and more.
These plates have flat bottom and can be transported for long distance if sealed by heat sealable film. These are produced and packed in Class 100000 clean workshop.
They have alphabetical sorting and corner cut marking for convenient tracking of samples. Each package has a separate article number/batch number identification, facilitating the quality tracking.
Certifications: Certified DNase/RNase and pyrogen free. Conformed to international SBS standards.
Product Family Options
Round wells
  • Désignation
    Volume
    Sterility
    Well Bottom Shape
    Packaging
  • Deep well plate, 48-well
    3.5 ml
    Non sterile
    U-bottom
    5/pack, 50/case
    Case of 50
    Case of 50 - $382.05
    $382.05
    Each
    Each - $382.05
    $382.05
    Case of 50
       $382.05
    Each
       $382.05
     : 504102
  • Deep well plate, 96-well
    0.4 ml
    Non sterile
    U-bottom
    5/pack, 50/case
    Case of 50
    Case of 50 - $327.79
    $327.79
    Each
    Each - $327.79
    $327.79
    Case of 50
       $327.79
    Each
       $327.79
     : 501102
  • Deep well plate, 96-well
    0.4 ml
    Sterile
    U-bottom
    5/pack, 50/case
    Case of 50
    Case of 50 - $363.51
    $363.51
    Each
    Each - $363.51
    $363.51
    Case of 50
       $363.51
    Each
       $363.51
     : 501162
  • Deep well plate, 96-well
    0.5 ml
    Non sterile
    V-bottom
    10/pack, 50/case
    Case of 50
    Case of 50 - $326.96
    $326.96
    Each
    Each - $326.96
    $326.96
    Case of 50
       $326.96
    Each
       $326.96
     : 501101
  • Deep well plate, 96-well
    0.5 ml
    Sterile
    V-bottom
    10/pack, 50/case
    Case of 50
    Case of 50 - $369.05
    $369.05
    Each
    Each - $369.05
    $369.05
    Case of 50
       $369.05
    Each
       $369.05
     : 501601
  • Deep well plate, 96-well
    1.0 ml
    Non sterile
    V-bottom
    5/pack, 50/case
    Case of 50
    Case of 50 - $326.96
    $326.96
    Each
    Each - $326.96
    $326.96
    Case of 50
       $326.96
    Each
       $326.96
     : 502101
  • Deep well plate, 96-well
    1.0 ml
    Non sterile
    U-bottom
    5/pack, 50/case
    Case of 50
    Case of 50 - $310.48
    $310.48
    Each
    Each - $310.48
    $310.48
    Case of 50
       $310.48
    Each
       $310.48
     : 502102
  • Deep well plate, 96-well
    1.0 ml
    Sterile
    U-bottom
    5/pack, 50/case
    Case of 50
    Case of 50 - $349.18
    $349.18
    Each
    Each - $349.18
    $349.18
    Case of 50
       $349.18
    Each
       $349.18
     : 502162
  • Deep well plate, 96-well
    1.0 ml
    Sterile
    V-bottom
    5/pack, 50/case
    Case of 50
    Case of 50 - $369.05
    $369.05
    Each
    Each - $369.05
    $369.05
    Case of 50
       $369.05
    Each
       $369.05
     : 502601
  • Deep well plate, 96-well
    1.3 ml
    Non sterile
    U-bottom
    5/pack, 50/case
    Case of 50
    Case of 50 - $276.99
    $276.99
    Each
    Each - $276.99
    $276.99
    Case of 50
       $276.99
    Each
       $276.99
     : 507001
  • Deep well plate, 96-well
    1.3 ml
    Sterile
    U-bottom
    5/pack, 50/case
    Case of 50
    Case of 50 - $276.99
    $276.99
    Each
    Each - $276.99
    $276.99
    Case of 50
       $276.99
    Each
       $276.99
     : 507101
  • Deep well plate, 96-well
    2.0 ml
    Non sterile
    U-bottom
    5/pack, 50/case
    Case of 50
    Case of 50 - $364.84
    $364.84
    Each
    Each - $364.84
    $364.84
    Case of 50
       $364.84
    Each
       $364.84
     : 503102
  • Deep well plate, 96-well
    2.0 ml
    Sterile
    U-bottom
    5/pack, 50/case
    Case of 50
    Case of 50 - $404.00
    $404.00
    Each
    Each - $404.00
    $404.00
    Case of 50
       $404.00
    Each
       $404.00
     : 503162
  • Deep well plate, 96-well
    0.36 ml
    Non sterile
    V-bottom
    5/pack, 50/case
    Case of 50
    Case of 50 - $327.79
    $327.79
    Each
    Each - $327.79
    $327.79
    Case of 50
       $327.79
    Each
       $327.79
     : 500101
  • Deep well plate, 96-well
    0.36 ml
    Sterile
    V-bottom
    5/pack, 50/case
    Case of 50
    Case of 50 - $363.51
    $363.51
    Each
    Each - $363.51
    $363.51
    Case of 50
       $363.51
    Each
       $363.51
     : 500161
Square wells
  • Désignation
    Volume
    Sterility
    Well Bottom Shape
    Packaging
  • Deep well plate, 24-well
    16 ml
    Sterile
    V-bottom
    1/pack, 25/case
    Case of 25
    Case of 25 - $186.22
    $186.22
    Each
    Each - $186.22
    $186.22
    Case of 25
       $186.22
    Each
       $186.22
     : 510001
  • Deep well plate, 48-well
    4.6 ml
    Non sterile
    U-bottom
    5/pack, 50/case
    Case of 50
    Case of 50 - $334.75
    $334.75
    Each
    Each - $334.75
    $334.75
    Case of 50
       $334.75
    Each
       $334.75
     : 504002
  • Deep well plate, 48-well
    4.6 ml
    Sterile
    U-bottom
    5/pack, 50/case
    Case of 50
    Case of 50 - $376.81
    $376.81
    Each
    Each - $376.81
    $376.81
    Case of 50
       $376.81
    Each
       $376.81
     : 504062
  • Deep well plate, 96-well
    0.5 ml
    Non sterile
    V-conical bottom
    5/pack, 50 /case
    Case of 50
    Case of 50 - $407.04
    $407.04
    Each
    Each - $407.04
    $407.04
    Case of 50
       $407.04
    Each
       $407.04
     : 500021
  • Deep well plate, 96-well
    0.5 ml
    Sterile
    V-conical bottom
    5/pack, 50/case
    Case of 50
    Case of 50 - $433.44
    $433.44
    Each
    Each - $433.44
    $433.44
    Case of 50
       $433.44
    Each
       $433.44
     : 500621
  • Deep well plate, 96-well
    1.0 ml
    Non sterile
    U-bottom
    5/pack, 50 /case
    Case of 50
    Case of 50 - $310.31
    $310.31
    Each
    Each - $310.31
    $310.31
    Case of 50
       $310.31
    Each
       $310.31
     : 501002
  • Deep well plate, 96-well
    1.0 ml
    Sterile
    U-bottom
    5/pack, 50 /case
    Case of 50
    Case of 50 - $349.47
    $349.47
    Each
    Each - $349.47
    $349.47
    Case of 50
       $349.47
    Each
       $349.47
     : 501062
  • Deep well plate, 96-well
    1.6 ml
    Non sterile
    U-bottom
    5/pack, 50 /case
    Case of 50
    Case of 50 - $307.62
    $307.62
    Each
    Each - $307.62
    $307.62
    Case of 50
       $307.62
    Each
       $307.62
     : 502002
  • Deep well plate, 96-well
    1.6 ml
    Sterile
    U-bottom
    5/pack, 50 /case
    Case of 50
    Case of 50 - $349.47
    $349.47
    Each
    Each - $349.47
    $349.47
    Case of 50
       $349.47
    Each
       $349.47
     : 502062
  • Deep well plate, 96-well
    2.2 ml
    Non sterile
    V-bottom
    5/pack, 50 /case
    Case of 50
    Case of 50 - $329.89
    $329.89
    Each
    Each - $329.89
    $329.89
    Case of 50
       $329.89
    Each
       $329.89
     : 503001
  • Deep well plate, 96-well
    2.2 ml
    Non sterile
    U-bottom
    5/pack, 50 /case
    Case of 50
    Case of 50 - $329.89
    $329.89
    Each
    Each - $329.89
    $329.89
    Case of 50
       $329.89
    Each
       $329.89
     : 503002
  • Deep well plate, 96-well
    2.2 ml
    Non sterile
    V-conical bottom
    5/pack, 50 /case
    Case of 50
    Case of 50 - $442.34
    $442.34
    Each
    Each - $442.34
    $442.34
    Case of 50
       $442.34
    Each
       $442.34
     : 503201
  • Deep well plate, 96-well
    2.2 ml
    Sterile
    U-bottom
    5/pack, 50/case
    Case of 50
    Case of 50 - $369.05
    $369.05
    Each
    Each - $369.05
    $369.05
    Case of 50
       $369.05
    Each
       $369.05
     : 503062
  • Deep well plate, 96-well
    2.2 ml
    Sterile
    V-bottom
    5/pack, 50 /case
    Case of 50
    Case of 50 - $369.05
    $369.05
    Each
    Each - $369.05
    $369.05
    Case of 50
       $369.05
    Each
       $369.05
     : 503501
  • Deep well plate, 96-well
    2.2 ml
    Sterile
    V-conical bottom
    5/pack, 50/case
    Case of 50
    Case of 50 - $480.19
    $480.19
    Each
    Each - $480.19
    $480.19
    Case of 50
       $480.19
    Each
       $480.19
     : 503261
  • Deep well plate, 96-well
    2.2 ml
    Non sterile
    I-shaped, U-bottom
    5/pack, 50/case
    Case of 50
    Case of 50 - $186.06
    $186.06
    Each
    Each - $186.06
    $186.06
    Case of 50
       $186.06
    Each
       $186.06
     : 503711
  • Deep well plate, 96-well
    2.2 ml
    Sterile
    I-shaped, U-bottom
    5/pack, 50/case
    Case of 50
    Case of 50 - $187.68
    $187.68
    Each
    Each - $187.68
    $187.68
    Case of 50
       $187.68
    Each
       $187.68
     : 503761
  • Deep well plate, 384-well
    120 µl
    Non sterile
    V-bottom
    5/pack, 50/case
    Case of 50
    Case of 50 - $451.57
    $451.57
    Each
    Each - $451.57
    $451.57
    Case of 50
       $451.57
    Each
       $451.57
     : 504701
Silicone sealing mats
  •  
    Sterility
    Désignation
    Packaging
    Plage de température
  • Non sterile
    Silicone sealing mat, round well, 96-well, 2 ml
    10/pack, 50/case
    −50...+200 °C
    Case of 50
    Case of 50 - $365.26
    $365.26
    Each
    Each - $365.26
    $365.26
    Case of 50
       $365.26
    Each
       $365.26
  •  
    Sterility
    Désignation
    Packaging
    Plage de température
  • Non sterile
    Silicone sealing mat, round well, 96-well, pre-slit, 2 ml
    10/pack, 50/case
    −50...+200 °C
    Case of 50
    Case of 50 - $365.26
    $365.26
    Each
    Each - $365.26
    $365.26
    Case of 50
       $365.26
    Each
       $365.26
  •  
    Sterility
    Désignation
    Packaging
    Plage de température
  • Sterile
    Silicone sealing mat, square well, 48-well
    10/pack, 50/case
    −50...+200 °C
    Case of 50
    Case of 50 - $407.24
    $407.24
    Each
    Each - $407.24
    $407.24
    Case of 50
       $407.24
    Each
       $407.24
  •  
    Sterility
    Désignation
    Packaging
    Plage de température
  • Non sterile
    Silicone sealing mat, square well, 48-well
    10/pack, 50/case
    −50...+200 °C
    Case of 50
    Case of 50 - $373.66
    $373.66
    Each
    Each - $373.66
    $373.66
    Case of 50
       $373.66
    Each
       $373.66
  •  
    Sterility
    Désignation
    Packaging
    Plage de température
  • Non sterile
    Silicone sealing mat, square well, 96-well
    10/pack, 50/case
    −50...+200 °C
    Case of 50
    Case of 50 - $365.26
    $365.26
    Each
    Each - $365.26
    $365.26
    Case of 50
       $365.26
    Each
       $365.26
  •  
    Sterility
    Désignation
    Packaging
    Plage de température
  • Non sterile
    Silicone sealing mat, square well, 96-well
    10/pack, 50/case
    −50...+200 °C
    Case of 50
    Case of 50 - $368.56
    $368.56
    Each
    Each - $368.56
    $368.56
    Case of 50
       $368.56
    Each
       $368.56
  •  
    Sterility
    Désignation
    Packaging
    Plage de température
  • Non sterile
    Silicone sealing mat, square well, 384-well
    10/pack, 50/case
    −50...+200 °C
    Case of 50
    Case of 50 - $373.66
    $373.66
    Each
    Each - $373.66
    $373.66
    Case of 50
       $373.66
    Each
       $373.66
  •  
    Sterility
    Désignation
    Packaging
    Plage de température
  • Non sterile
    Silicone sealing mat, square well, 24-well
    10/pack, 50/case
    −50...+200 °C
    Case of 50
    Case of 50 - $373.66
    $373.66
    Each
    Each - $373.66
    $373.66
    Case of 50
       $373.66
    Each
       $373.66
  •  
    Sterility
    Désignation
    Packaging
    Plage de température
  • Non sterile
    Silicone sealing mat, round well, 96-well, can be punctured, 1 ml
    10/pack, 50/case
    −50...+200 °C
    Case of 50
    Case of 50 - $365.26
    $365.26
    Each
    Each - $365.26
    $365.26
    Case of 50
       $365.26
    Each
       $365.26
Plastic comb
  •  
    Désignation
  • Plastic comb, 24 tip, sterile, compatible with Thermo KingFisher System, 1/pack, 25/case
    Case of 25
    Case of 25 - $255.15
    $255.15
    Each
    Each - $255.15
    $255.15
    Case of 25
       $255.15
    Each
       $255.15
  •  
    Désignation
  • Plastic comb, 8-strip, non sterile, compatible with 503711 and 503761, 2/pack, 50/box, 500/case
    Case of 500
    Case of 500 - $665.29
    $665.29
    Each
    Each - $665.29
    $665.29
    Case of 500
       $665.29
    Each
       $665.29
  •  
    Désignation
  • Plastic comb, 8-strip, sterile, compatible with 503711 and 503761, 2/pack, 50/box, 500/case
    Case of 500
    Case of 500 - $712.19
    $712.19
    Each
    Each - $712.19
    $712.19
    Case of 500
       $712.19
    Each
       $712.19
  •  
    Désignation
  • Combs for magnetic applications, 96 tip, sterile, 2/pack, 50/case
    Case of 50
    Case of 50 - $480.19
    $480.19
    Each
    Each - $480.19
    $480.19
    Case of 50
       $480.19
    Each
       $480.19
  •  
    Désignation
  • Combs for magnetic applications, 96 tip, non sterile, 2/pack, 50/case
    Case of 50
    Case of 50 - $451.27
    $451.27
    Each
    Each - $451.27
    $451.27
    Case of 50
       $451.27
    Each
       $451.27