Cell Culture Plates | Avantor
Cell Culture Plates
Supplier:   NEST SCIENTIFIC
Cell Culture Plates
Supplier:  NEST SCIENTIFIC
Certificates
Cell Culture Plates
36 Options Available Below
Product Details & Documents
NEST cell culture plates have round edges for better handling. Low profile, low-evaporation lid design assures uniform growth.
  • Sterilized by E-beam, SAL=10⁻⁶
  • High clarity, 100% virgin polystyrene
  • TC refers to vacuum plasma tissue culture treatment
  • Clear lot number for batch traceability
  • Markings of well coordinates available for 12, 24, 48 and 384 well plates
  • Non-pyrogenic, DNase/Rnase free
Tissue treated and non-treated are both available. Non-treated growth surface is more hydrophobic than tissue culture surface and are similar to the surface of a bacteriological Petri dish. Small feet on the bottom corners of each plate eliminate scraping of the microscope lens.
Grooves are designed at the edge of the culture plate to avoid the edge effect to the greatest extent and to ensure that the cells maintain their optimal state during cell culture.
Product Family Options
Individually packaged in peelable tyvek film and blister
Informations sur le produit
  • Désignation
    Nb de puits
    Matière
    Working Volume
    Cell Growth Area
    Color
    Sterility
    Surface Treatment
    Packaging
    Availability
    Price
  • Cell culture plate, TC-treated, flat bottom
    6
    Polystyrene
    1.9 - 2.9 ml
    9.5 cm²
    Clear
    Sterile
    Tissue culture (TC)
    1/pack, 50/case
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $187.68
    $187.68
    Log in pour la disponibilité
    Each
    Each - $187.68
    $187.68
    Case of 50
       $187.68
    Each
       $187.68
    Supplier Number: 703001
    Spécifications:
    More
    Specifications
    Cat. No.
    77508-532
  • Cell culture plate, non-treated, flat bottom
    6
    Polystyrene
    1.9 - 2.9 ml
    9.5 cm²
    Clear
    Sterile
    Non-treated
    1/pack, 50/case
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $182.66
    $182.66
    Log in pour la disponibilité
    Each
    Each - $182.66
    $182.66
    Case of 50
       $182.66
    Each
       $182.66
    Supplier Number: 703011
    Spécifications:
    More
    Specifications
    Cat. No.
    77508-538
  • Low evap EDGE plate, non-treated, flat bottom
    6
    Polystyrene
    1.9 - 2.9 ml
    9.5 cm²
    Clear
    Sterile
    Non-treated
    1/pack, 50/case
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $283.97
    $283.97
    Log in pour la disponibilité
    Each
    Each - $283.97
    $283.97
    Case of 50
       $283.97
    Each
       $283.97
    Supplier Number: 714001
    Spécifications:
    More
    Specifications
    Cat. No.
    77508-598
  • Low evap EDGE plate, TC-treated, flat bottom
    6
    Polystyrene
    1.9 - 2.9 ml
    9.5 cm²
    Clear
    Sterile
    Tissue culture (TC)
    1/pack, 50/case
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $286.54
    $286.54
    Log in pour la disponibilité
    Each
    Each - $286.54
    $286.54
    Case of 50
       $286.54
    Each
       $286.54
    Supplier Number: 714011
    Spécifications:
    More
    Specifications
    Cat. No.
    77508-600
  • Cell culture plate, TC-treated, flat bottom
    12
    Polystyrene
    0.76 - 1.14 ml
    3.6 cm²
    Clear
    Sterile
    Tissue culture (TC)
    1/pack, 50/case
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $199.81
    $199.81
    Log in pour la disponibilité
    Each
    Each - $199.81
    $199.81
    Case of 50
       $199.81
    Each
       $199.81
    Supplier Number: 712001
    Spécifications:
    More
    Specifications
    Cat. No.
    77508-586
  • Cell culture plate, non-treated, flat bottom
    12
    Polystyrene
    0.76 - 1.14 ml
    3.6 cm²
    Clear
    Sterile
    Non-treated
    1/pack, 50/case
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $193.51
    $193.51
    Log in pour la disponibilité
    Each
    Each - $193.51
    $193.51
    Case of 50
       $193.51
    Each
       $193.51
    Supplier Number: 712011
    Spécifications:
    More
    Specifications
    Cat. No.
    77508-592
  • Cell culture plate, TC-treated, flat bottom
    24
    Polystyrene
    0.38 - 0.57 ml
    1.9 cm²
    Clear
    Sterile
    Tissue culture (TC)
    1/pack, 50/case
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $199.17
    $199.17
    Log in pour la disponibilité
    Each
    Each - $199.17
    $199.17
    Case of 50
       $199.17
    Each
       $199.17
    Supplier Number: 702001
    Spécifications:
    More
    Specifications
    Cat. No.
    77508-524
  • Cell culture plate, non-treated, flat bottom
    24
    Polystyrene
    0.38 - 0.57 ml
    1.9 cm²
    Clear
    Sterile
    Non-treated
    1/pack, 50/case
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $194.15
    $194.15
    Log in pour la disponibilité
    Each
    Each - $194.15
    $194.15
    Case of 50
       $194.15
    Each
       $194.15
    Supplier Number: 702011
    Spécifications:
    More
    Specifications
    Cat. No.
    77508-530
  • Cell culture plate, TC-treated, flat bottom
    48
    Polystyrene
    0.19 - 0.285 ml
    0.88 cm²
    Clear
    Sterile
    Tissue culture (TC)
    1/pack, 50/case
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $203.53
    $203.53
    Log in pour la disponibilité
    Each
    Each - $203.53
    $203.53
    Case of 50
       $203.53
    Each
       $203.53
    Supplier Number: 748001
    Spécifications:
    More
    Specifications
    Cat. No.
    77508-730
  • Cell culture plate, non-treated, flat bottom
    48
    Polystyrene
    0.19 - 0.285 ml
    0.88 cm²
    Clear
    Sterile
    Non-treated
    1/pack, 50/case
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $194.15
    $194.15
    Log in pour la disponibilité
    Each
    Each - $194.15
    $194.15
    Case of 50
       $194.15
    Each
       $194.15
    Supplier Number: 748011
    Spécifications:
    More
    Specifications
    Cat. No.
    77508-736
  • Cell culture plate, TC-treated, flat bottom
    96
    Polystyrene
    0.1 - 0.2 ml
    0.32 cm²
    Clear
    Sterile
    Tissue culture (TC)
    1/pack, 100/case
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 100
    Case of 100 - $359.66
    $359.66
    Log in pour la disponibilité
    Each
    Each - $359.66
    $359.66
    Case of 100
       $359.66
    Each
       $359.66
    Supplier Number: 701001
    Spécifications:
    More
    Specifications
    Cat. No.
    77508-504
  • Cell culture plate, non-treated, flat bottom
    96
    Polystyrene
    0.1 - 0.2 ml
    0.32 cm²
    Clear
    Sterile
    Non-treated
    1/pack, 100/case
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 100
    Case of 100 - $342.19
    $342.19
    Log in pour la disponibilité
    Each
    Each - $342.19
    $342.19
    Case of 100
       $342.19
    Each
       $342.19
    Supplier Number: 701011
    Spécifications:
    More
    Specifications
    Cat. No.
    77508-510
  • Cell culture plate, TC-treated, U-bottom
    96
    Polystyrene
    0.1 - 0.2 ml
    0.66 cm²
    Clear
    Sterile
    Tissue culture (TC)
    1/pack, 100/case
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 100
    Case of 100 - $389.27
    $389.27
    Log in pour la disponibilité
    Each
    Each - $389.27
    $389.27
    Case of 100
       $389.27
    Each
       $389.27
    Supplier Number: 701101
    Spécifications:
    More
    Specifications
    Cat. No.
    77508-512
  • Cell culture plate, non-treated, U-bottom
    96
    Polystyrene
    0.1 - 0.2 ml
    0.66 cm²
    Clear
    Sterile
    Non-treated
    1/pack, 100/case
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 100
    Case of 100 - $382.97
    $382.97
    Log in pour la disponibilité
    Each
    Each - $382.97
    $382.97
    Case of 100
       $382.97
    Each
       $382.97
    Supplier Number: 701111
    Spécifications:
    More
    Specifications
    Cat. No.
    77508-514
  • Cell culture plate, TC-treated, V-bottom
    96
    Polystyrene
    0.1 - 0.2 ml
    0.41 cm²
    Clear
    Sterile
    Tissue culture (TC)
    1/pack, 100/case
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 100
    Case of 100 - $389.27
    $389.27
    Log in pour la disponibilité
    Each
    Each - $389.27
    $389.27
    Case of 100
       $389.27
    Each
       $389.27
    Supplier Number: 701201
    Spécifications:
    More
    Specifications
    Cat. No.
    77508-516
  • Cell culture plate, non-treated, V-bottom
    96
    Polystyrene
    0.1 - 0.2 ml
    0.41 cm²
    Clear
    Sterile
    Non-treated
    1/pack, 100/case
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 100
    Case of 100 - $382.97
    $382.97
    Log in pour la disponibilité
    Each
    Each - $382.97
    $382.97
    Case of 100
       $382.97
    Each
       $382.97
    Supplier Number: 701211
    Spécifications:
    More
    Specifications
    Cat. No.
    77508-518
  • Cell culture plate, TC-treated, flat bottom
    96
    Polystyrene
    0.1 - 0.2 ml
    0.32 cm²
    White
    Sterile
    Tissue culture (TC)
    1/pack, 100/case
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 100
    Case of 100 - $464.02
    $464.02
    Log in pour la disponibilité
    Each
    Each - $464.02
    $464.02
    Case of 100
       $464.02
    Each
       $464.02
    Supplier Number: 701301
    Spécifications:
    More
    Specifications
    Cat. No.
    77508-520
  • Cell culture plate, non-treated, flat bottom
    96
    Polystyrene
    0.1 - 0.2 ml
    0.32 cm²
    White
    Sterile
    Non-treated
    1/pack, 100/case
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 100
    Case of 100 - $451.69
    $451.69
    Log in pour la disponibilité
    Each
    Each - $451.69
    $451.69
    Case of 100
       $451.69
    Each
       $451.69
    Supplier Number: 701311
    Spécifications:
    More
    Specifications
    Cat. No.
    77508-522
  • Low evap EDGE plate, non-treated, flat bottom
    96
    Polystyrene
    0.1 - 0.2 ml
    0.32 cm²
    Clear
    Sterile
    Non-treated
    1/pack, 100/case
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $493.25
    $493.25
    Log in pour la disponibilité
    Each
    Each - $493.25
    $493.25
    Case of 50
       $493.25
    Each
       $493.25
    Supplier Number: 713001
    Spécifications:
    More
    Specifications
    Cat. No.
    77508-594
  • Low evap EDGE plate, TC-treated, flat bottom
    96
    Polystyrene
    0.1 - 0.2 ml
    0.32 cm²
    Clear
    Sterile
    Tissue culture (TC)
    1/pack, 100/case
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 100
    Case of 100 - $517.21
    $517.21
    Log in pour la disponibilité
    Each
    Each - $517.21
    $517.21
    Case of 100
       $517.21
    Each
       $517.21
    Supplier Number: 713011
    Spécifications:
    More
    Specifications
    Cat. No.
    77508-596
  • Cell culture plate, TC-treated, flat bottom
    384
    Polystyrene
    0.025 - 0.05 ml
    0.11 cm²
    Clear
    Sterile
    Tissue culture (TC)
    1/pack, 100/case
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 100
    Case of 100 - $831.78
    $831.78
    Log in pour la disponibilité
    Each
    Each - $831.78
    $831.78
    Case of 100
       $831.78
    Each
       $831.78
    Supplier Number: 761001
    Spécifications:
    More
    Specifications
    Cat. No.
    77508-754
  • Cell culture plate, non-treated, flat bottom
    384
    Polystyrene
    0.025 - 0.05 ml
    0.11 cm²
    Clear
    Sterile
    Non-treated
    1/pack, 100/case
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 100
    Case of 100 - $756.33
    $756.33
    Log in pour la disponibilité
    Each
    Each - $756.33
    $756.33
    Case of 100
       $756.33
    Each
       $756.33
    Supplier Number: 761011
    Spécifications:
    More
    Specifications
    Cat. No.
    77508-756
  • Cell culture plate, TC-treated, flat bottom
    384
    Polystyrene
    0.025 - 0.05 ml
    0.11 cm²
    Black
    Sterile
    Tissue culture (TC)
    1/pack, 100/case
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 100
    Case of 100 - $831.78
    $831.78
    Log in pour la disponibilité
    Each
    Each - $831.78
    $831.78
    Case of 100
       $831.78
    Each
       $831.78
    Supplier Number: 761301
    Spécifications:
    More
    Specifications
    Cat. No.
    77508-758
  • Cell culture plate, non-treated, flat bottom
    384
    Polystyrene
    0.025 - 0.05 ml
    0.11 cm²
    Black
    Sterile
    Non-treated
    1/pack, 100/case
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 100
    Case of 100 - $756.33
    $756.33
    Log in pour la disponibilité
    Each
    Each - $756.33
    $756.33
    Case of 100
       $756.33
    Each
       $756.33
    Supplier Number: 761311
    Spécifications:
    More
    Specifications
    Cat. No.
    77508-760
  • Cell culture plate, TC-treated, flat bottom
    384
    Polystyrene
    0.025 - 0.05 ml
    0.11 cm²
    White
    Sterile
    Tissue culture (TC)
    1/pack, 100/case
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 100
    Case of 100 - $839.37
    $839.37
    Log in pour la disponibilité
    Each
    Each - $839.37
    $839.37
    Case of 100
       $839.37
    Each
       $839.37
    Supplier Number: 761601
    Spécifications:
    More
    Specifications
    Cat. No.
    77508-762
  • Cell culture plate, non-treated, flat bottom
    384
    Polystyrene
    0.025 - 0.05 ml
    0.11 cm²
    White
    Sterile
    Non-treated
    1/pack, 100/case
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 100
    Case of 100 - $756.33
    $756.33
    Log in pour la disponibilité
    Each
    Each - $756.33
    $756.33
    Case of 100
       $756.33
    Each
       $756.33
    Supplier Number: 761611
    Spécifications:
    More
    Specifications
    Cat. No.
    77508-764
Plastic bag package, 10/pack
Informations sur le produit
  • Désignation
    Nb de puits
    Matière
    Working Volume
    Cell Growth Area
    Color
    Sterility
    Surface Treatment
    Packaging
    Availability
    Price
  • Cell culture plate, TC-treated, flat bottom
    6
    Polystyrene
    1.9 - 2.9 ml
    9.5 cm²
    Clear
    Sterile
    Tissue culture (TC)
    10/pack, 50/case
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $156.46
    $156.46
    Log in pour la disponibilité
    Each
    Each - $156.46
    $156.46
    Case of 50
       $156.46
    Each
       $156.46
    Supplier Number: 703002
    Spécifications:
    More
    Specifications
    Cat. No.
    77508-534
  • Cell culture plate, TC-treated, flat bottom
    12
    Polystyrene
    0.76 - 1.14 ml
    3.6 cm²
    Clear
    Sterile
    Tissue culture (TC)
    10/pack, 50/case
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $165.19
    $165.19
    Log in pour la disponibilité
    Each
    Each - $165.19
    $165.19
    Case of 50
       $165.19
    Each
       $165.19
    Supplier Number: 712002
    Spécifications:
    More
    Specifications
    Cat. No.
    77508-588
  • Cell culture plate, TC-treated, flat bottom
    24
    Polystyrene
    0.38 - 0.57 ml
    1.9 cm²
    Clear
    Sterile
    Tissue culture (TC)
    10/pack, 50/case
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $165.19
    $165.19
    Log in pour la disponibilité
    Each
    Each - $165.19
    $165.19
    Case of 50
       $165.19
    Each
       $165.19
    Supplier Number: 702002
    Spécifications:
    More
    Specifications
    Cat. No.
    77508-526
  • Cell culture plate, TC-treated, flat bottom
    48
    Polystyrene
    0.19 - 0.285 ml
    0.88 cm²
    Clear
    Sterile
    Tissue culture (TC)
    10/pack, 50/case
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $170.53
    $170.53
    Log in pour la disponibilité
    Each
    Each - $170.53
    $170.53
    Case of 50
       $170.53
    Each
       $170.53
    Supplier Number: 748002
    Spécifications:
    More
    Specifications
    Cat. No.
    77508-732
  • Cell culture plate, TC-treated, flat bottom
    96
    Polystyrene
    0.1 - 0.2 ml
    0.32 cm²
    Clear
    Sterile
    Tissue culture (TC)
    10/pack, 100/case
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 100
    Case of 100 - $316.95
    $316.95
    Log in pour la disponibilité
    Each
    Each - $316.95
    $316.95
    Case of 100
       $316.95
    Each
       $316.95
    Supplier Number: 701002
    Spécifications:
    More
    Specifications
    Cat. No.
    77508-506
Plastic bag package, 1/pack
Informations sur le produit
  • Désignation
    Nb de puits
    Matière
    Working Volume
    Cell Growth Area
    Color
    Sterility
    Surface Treatment
    Packaging
    Availability
    Price
  • Cell culture plate, TC-treated, flat bottom
    6
    Polystyrene
    1.9 - 2.9 ml
    9.5 cm²
    Clear
    Sterile
    Tissue culture (TC)
    1/pack, 50/case
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $162.93
    $162.93
    Log in pour la disponibilité
    Each
    Each - $162.93
    $162.93
    Case of 50
       $162.93
    Each
       $162.93
    Supplier Number: 703003
    Spécifications:
    More
    Specifications
    Cat. No.
    77508-536
  • Cell culture plate, TC-treated, flat bottom
    12
    Polystyrene
    0.76 - 1.14 ml
    3.6 cm²
    Clear
    Sterile
    Tissue culture (TC)
    1/pack, 50/case
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $173.12
    $173.12
    Log in pour la disponibilité
    Each
    Each - $173.12
    $173.12
    Case of 50
       $173.12
    Each
       $173.12
    Supplier Number: 712003
    Spécifications:
    More
    Specifications
    Cat. No.
    77508-590
  • Cell culture plate, TC-treated, flat bottom
    24
    Polystyrene
    0.38 - 0.57 ml
    1.9 cm²
    Clear
    Sterile
    Tissue culture (TC)
    1/pack, 50/case
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $173.12
    $173.12
    Log in pour la disponibilité
    Each
    Each - $173.12
    $173.12
    Case of 50
       $173.12
    Each
       $173.12
    Supplier Number: 702003
    Spécifications:
    More
    Specifications
    Cat. No.
    77508-528
  • Cell culture plate, TC-treated, flat bottom
    48
    Polystyrene
    0.19 - 0.285 ml
    0.88 cm²
    Clear
    Sterile
    Tissue culture (TC)
    1/pack, 50/case
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $176.35
    $176.35
    Log in pour la disponibilité
    Each
    Each - $176.35
    $176.35
    Case of 50
       $176.35
    Each
       $176.35
    Supplier Number: 748003
    Spécifications:
    More
    Specifications
    Cat. No.
    77508-734
  • Cell culture plate, TC-treated, flat bottom
    96
    Polystyrene
    0.1 - 0.2 ml
    0.32 cm²
    Clear
    Sterile
    Tissue culture (TC)
    1/pack, 100/case
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 100
    Case of 100 - $322.25
    $322.25
    Log in pour la disponibilité
    Each
    Each - $322.25
    $322.25
    Case of 100
       $322.25
    Each
       $322.25
    Supplier Number: 701003
    Spécifications:
    More
    Specifications
    Cat. No.
    77508-508