Colored Cap Inserts
Colored Cap Inserts
Colored Cap Inserts
Six colored cap inserts for easy identification. Compatible with regular cryogenic vials that with external thread caps only.
 : 100/pack; 1000/case
Product Family Options
  • Désignation
    Color
  • Coloured cap insert
    Blue
     : 611004
  • Coloured cap insert
    Green
    Case of 1,000
    Case of 1,000 - $98.52
    $98.52
    Each
    Each - $98.52
    $98.52
    Case of 1,000
       $98.52
    Each
       $98.52
     : 611005
  • Coloured cap insert
    Purple
     : 611006
  • Coloured cap insert
    Red
     : 611002
  • Coloured cap insert
    White
     : 611001
  • Coloured cap insert
    Yellow
    Case of 1,000
    Case of 1,000 - $98.30
    $98.30
    Each
    Each - $98.30
    $98.30
    Case of 1,000
       $98.30
    Each
       $98.30
     : 611003