ULTRA-TURRAX® T 50 basic Laboratory/Pilot Scale Disperser/Homogenizer Kit, IKA® Works
Catalogue # 33998-030
Fournisseur: IKA Works
Spécifications
- Ø:45 mm
- Description:S 50 N - G 45 G, Coarse
- Cat. no.:33998-030
- Volume Range:0.5 - 20 ml
- Supplier No.:8003000
Spécifications
À propos de cet article
The T 50 basic is a high-efficiency dispersing instrument suited for a wide variety of applications
Variable speed drive is designed for continuous operation. Equipped with an interface connection for revolution counter, and an extension arm.
Informations sur les accessoires: Telescopic stand has a stable 46Wx42Dcm (181/8x161/2") H-shaped base to prevent tipping and a 3.4cm (111/32") diameter pneumatic spring stand rod to accommodate heavy attachments. Stand can support up to 10kg (22lbs.).
Ordering information: Manufacturer's two-year warranty on parts and labor. Dispersing elements are ordered separately. Disperser/homogenizer kit 82017-870 includes T 50 disperser, R 271 boss head clamp, R 2723 telescopic stand, and RH 5 strap clamp.