Robotic Pipette Tips
Supplier: NEST SCIENTIFIC
Jump to:
À propos de cet article
NEST robotic conductive tip can be applied in R and D of genomics, proteomics, cytomics, immunoassays, metabolomics, biopharmaceuticals and other high flux liquid transfer.
- Made of polymer material polypropylene (PP)
- Offers high efficiency and repeatability
- With barcode
Surface owns high hydrophobicity as treated with a special technique making for certain purposes.
12 Options Available Below
Product Details & Documents
NEST robotic conductive tip can be applied in R and D of genomics, proteomics, cytomics, immunoassays, metabolomics, biopharmaceuticals and other high flux liquid transfer.
- Made of polymer material polypropylene (PP)
- Offers high efficiency and repeatability
- With barcode
Surface owns high hydrophobicity as treated with a special technique making for certain purposes.
Product Family Options
Filtered tips
Informations sur le produit
- VolumeSterilePackedAvailabilityPrice
- 50 µl+96 pieces/rack 24 racks/packCase of 2,304$404.79Each$404.79Supplier Number: 345513
- 50 µl+96 pieces/box, 10 boxes/pack, 5 packs/caseCase of 4,800$648.30Each$648.30Supplier Number: 345519
- 300 µl+96 pieces/rack 24 racks/packCase of 2,304$444.01Each$444.01Supplier Number: 345613
- 300 µl+96 pieces/box, 10 boxes/pack, 5 packs/caseCase of 4,800$695.71Each$695.71Supplier Number: 345619
- 1000 µl+96 pieces/rack 24 racks/packCase of 2,304$509.55Each$509.55Supplier Number: 345713
- 1000 µl+96 pieces/box, 10 boxes/pack, 5 packs/caseCase of 4,800$841.41Each$841.41Supplier Number: 345719
Non filtered tips
Informations sur le produit
- VolumeSterilePackedAvailabilityPrice
- 50 µl+96 pieces/rack 24 racks/packCase of 2,304$339.76Each$339.76Supplier Number: 345503
- 50 µl+96 pieces/box, 10 boxes/pack, 5 packs/caseCase of 4,800$462.03Each$462.03Supplier Number: 345509
- 300 µl+96 pieces/rack 24 racks/packCase of 2,304$365.57Each$365.57Supplier Number: 345603
- 300 µl+96 pieces/box, 10 boxes/pack, 5 packs/caseCase of 4,800$575.39Each$575.39Supplier Number: 345609
- 1000 µl+96 pieces/rack 24 racks/packCase of 2,304$417.89Each$417.89Supplier Number: 345703
- 1000 µl+96 pieces/box, 10 boxes/pack, 5 packs/caseCase of 4,800$670.30Each$670.30Supplier Number: 345709