Adjustable Wrist Grounding Strap, STATICO
Adjustable Wrist Grounding Strap, STATICO
Supplier:   Statico
Adjustable Wrist Grounding Strap, STATICO
Supplier:  Statico
Certificates
Adjustable Wrist Grounding Strap, STATICO
Adjustable Wrist Grounding Strap, STATICO
3 Options Available Below
Product Details & Documents
Adjustable light blue wrist grounding strap with 360⁰ skin contact
A 1 Meg current limiting resistor is molded-in at the snap end termination of the cord
Certifications: Meets ANSI/ESD S1.1-2013 & ANSI/ESD S20.20-2014.
Ordering information: Available with either 1.8 m (6') or 3.6 m (12') Coil Cord and an Alligator Clip.
Product Family Options
Informations sur le produit
  • Désignation
    Cord Length
    Availability
    Price
  • Wrist strap with coil cord and alligator clip
    1.8 m (6')
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $87.63
    $87.63
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $229.10
    $229.10
    Each
       $87.63
    Pack of 10
       $229.10
    Supplier Number: S0101
  • Wrist strap with coil cord and alligator clip
    1.8 m (6')
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $28.84
    $28.84
    Each
       $28.84
    Supplier Number: S0101
  • Wrist strap with coil cord and alligator clip
    3.6 m (12')
    Discontinued
    Supplier Number: -
  • Wrist strap (band) only
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $53.06
    $53.06
    Log in pour la disponibilité
    Pack of 20
    Pack of 20 - $1,061.20
    $1,061.20
    Each
       $53.06
    Pack of 20
       $1,061.20
    Supplier Number: S0101B