ELISA Plates
ELISA Plates
Supplier:   NEST SCIENTIFIC
ELISA Plates
Supplier:  NEST SCIENTIFIC
Certificates
4 Options Available Below
Product Details & Documents
NEST ELISA plate, made of polystyrene, is a safe, reliable, and effective supporter in enzyme-linked immuno-absorbent assay (ELISA). The features of the plates are crucial for the adsorption of antigens, antibodies, and antigen-antibody complexes.
  • Uniform thickness and size
  • High batch-to-batch stability and low coefficient of variation (CV) value
  • Both detachable and non-detachable flat-bottom plates are offered for customers to choose from based on their applications
  • The letters and numbers on the frame of the plates are convenient for detection and identification in experiments
  • High transparency of clear plate bottom
  • Greatly improves the flexibility in diagnostic testing (Only for detachable ELISA plates)
  • Each well is locked in the frame, all at the same horizontal (Only for detachable ELISA plates)
  • The design of 8-well strip allows for uniform reading and washing (Only for detachable ELISA plates)
ELISA plates from NEST adopts a unique surface treatment process, allowing to obtain ideal results in ELISA tests which are extensively employed in vitro diagnostics in medical and clinical laboratories.
The plates are made of polystyrene and are designed specifically for ELISAs. They present good adsorption performance and low baseline value.
White plates are best suited for luminescence measurements as they reflect the signal instead of absorbing it and hence enhance the luminescence signals. Raw materials show good light-proof properties, which greatly reduce well-to-well signal interference.
Product Family Options
Informations sur le produit
  • Désignation
    Nb de puits
    Matière
    Color
    Packaging
    Availability
    Price
  • High binding ELISA plate, detachable
    96
    PS
    White frame and transparent well
    5 plates/pack, 50 plates/case
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $195.29
    $195.29
    Log in pour la disponibilité
    Each
    Each - $195.29
    $195.29
    Case of 50
       $195.29
    Each
       $195.29
    Supplier Number: 504201
  • High binding ELISA plate, detachable
    96
    PS
    White frame and black well
    5 plates/pack, 50 plates/case
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $133.43
    $133.43
    Log in pour la disponibilité
    Each
    Each - $133.43
    $133.43
    Case of 50
       $133.43
    Each
       $133.43
    Supplier Number: 504261
  • High binding ELISA plate, detachable
    96
    PS
    White frame and white well
    5 plates/pack, 50 plates/case
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $268.15
    $268.15
    Log in pour la disponibilité
    Each
    Each - $268.15
    $268.15
    Case of 50
       $268.15
    Each
       $268.15
    Supplier Number: 504271
  • High binding ELISA plate, undetachable
    96
    PS
    Transparent
    5 plates/pack, 50 plates/case
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $156.89
    $156.89
    Log in pour la disponibilité
    Each
    Each - $156.89
    $156.89
    Case of 50
       $156.89
    Each
       $156.89
    Supplier Number: 514201