Cryogenic Vials, External or Internal Threaded
À propos de cet article
Cryogenic Vials are designed for convenient and secure storage. These vials are ideal for storing valuable biological samples, ensuring their integrity and viability for long-term preservation.
- Made of USP VI polypropylene
- Autoclave at 121 °C/15 lbs.
- Non-pyrogenic, DNase/RNase and human DNA free
- Sterilized by E-beam, SAL=10⁻⁶
- Clear graduations and white marking area
- Stored in gas-phase liquid nitrogen
External thread cap: Special screw cap designed without silicone ring to avoid risk of contamination.
Internal thread cap: Specialscrew cap designed with silicone O-ring to secure the sealing under various conditon.
23 Options Available Below
Product Details & Documents
Cryogenic Vials are designed for convenient and secure storage. These vials are ideal for storing valuable biological samples, ensuring their integrity and viability for long-term preservation.
- Made of USP VI polypropylene
- Autoclave at 121 °C/15 lbs.
- Non-pyrogenic, DNase/RNase and human DNA free
- Sterilized by E-beam, SAL=10⁻⁶
- Clear graduations and white marking area
- Stored in gas-phase liquid nitrogen
External thread cap: Special screw cap designed without silicone ring to avoid risk of contamination.
Internal thread cap: Specialscrew cap designed with silicone O-ring to secure the sealing under various conditon.
Documents
Product Family Options
External threaded
Informations sur le produit
- CapacitéBottom StyleMatièreAvec bouchonCap TypeSterilityPackagingAvailabilityPrice
- 0.5 mlRound bottom, self-standingPPOuiScrew capSterile50/pack, 500/boxDiscontinued-Supplier Number: -
- 0.5 mlRound bottom, self-standingPPOuiScrew capSterile9×9 racked packageCase of 1,296$747.42Each$747.42Supplier Number: 618003
- 1.2 mlRound bottom, self-standingPPOuiScrew capSterile50/pack, 500/boxDiscontinued-Supplier Number: 606001
- 1.2 mlRound bottom, self-standingPPOuiScrew capSterile9×9 racked packageCase of 1,134$693.15Each$693.15Supplier Number: 606003
- 2.0 mlRound bottom, self-standingPPOuiScrew capSterile50/pack, 500/boxCase of 2,000$1,052.46Each$1,052.46Supplier Number: 607001
- 2.0 mlRound bottom, self-standingPPOuiScrew capSterile9×9 racked packageCase of 972$822.58Each$822.58Supplier Number: 607003
- 3.0 mlRound bottom, self-standingPPOuiScrew capSterile50/pack, 500/boxCase of 1,000$531.05Each$531.05Supplier Number: 624001
- 3.0 mlRound bottom, self-standingPPOuiScrew capSterile9×9 racked packageCase of 648$850.81Each$850.81Supplier Number: 624003
- 4.0 mlRound bottom, self-standingPPOuiScrew capSterile50/pack, 500/boxDiscontinued-Supplier Number: -
- 4.0 mlRound bottom, self-standingPPOuiScrew capSterile9×9 racked packageCase of 648$836.40Each$836.40Supplier Number: 608003
- 5.0 mlRound bottom, self-standingPPOuiScrew capSterile50/pack, 500/boxCase of 1,000$727.54Each$727.54Supplier Number: 609001
- 5.0 mlRound bottom, self-standingPPOuiScrew capSterile9×9 racked packageCase of 486$519.88Each$519.88Supplier Number: 609003
Internal threaded
Informations sur le produit
- CapacitéBottom StyleMatièreAvec bouchonCap TypeSterilityPackagingAvailabilityPrice
- 0.5 mlRound bottom, self-standingPPOuiScrew capSterile50/pack, 500/boxCase of 2,000$877.06Each$877.06Supplier Number: 618101
- 0.5 mlRound bottom, self-standingPPOuiScrew capSterile10×10 racked packageCase of 1,600$922.74Each$922.74Supplier Number: 618102
- 0.5 mlRound bottom, self-standingPPOuiScrew capSterile9×9 racked packageCase of 1,296$890.69Each$890.69Supplier Number: 618103
- 1.2 mlRound bottom, self-standingPPOuiScrew capSterile50/pack, 500/boxDiscontinued-Supplier Number: 606101
- 1.2 mlRound bottom, self-standingPPOuiScrew capSterile10×10 racked packageCase of 1,400$891.89Each$891.89Supplier Number: 606102
- 1.2 mlRound bottom, self-standingPPOuiScrew capSterile9×9 racked packageCase of 1,134$855.13Each$855.13Supplier Number: 606103
- 2.0 mlRound bottom, self-standingPPOuiScrew capSterile50/pack, 500/boxCase of 2,000$1,112.65Each$1,112.65Supplier Number: 607101
- 2.0 mlRound bottom, self-standingPPOuiScrew capSterile10×10 racked packageCase of 1,200$940.03Each$940.03Supplier Number: 607102
- 2.0 mlRound bottom, self-standingPPOuiScrew capSterile9×9 racked packageCase of 972$1,009.76Each$1,009.76Supplier Number: 607103
- 3.0 mlRound bottom, self-standingPPOuiScrew capSterile50/pack, 500/boxCase of 1,000$577.23Each$577.23Supplier Number: 624101
- 3.0 mlRound bottom, self-standingPPOuiScrew capSterile9×9 racked packageCase of 648$565.56Each$565.56Supplier Number: 624103
- 4.0 mlRound bottom, self-standingPPOuiScrew capSterile50/pack, 500/boxCase of 1,000$568.33Each$568.33Supplier Number: 608101
- 4.0 mlRound bottom, self-standingPPOuiScrew capSterile9×9 racked packageCase of 648$580.09Each$580.09Supplier Number: 608103
- 5.0 mlRound bottom, self-standingPPOuiScrew capSterile50/pack, 500/boxCase of 1,000$786.02Each$786.02Supplier Number: 609101
- 5.0 mlRound bottom, self-standingPPOuiScrew capSterile9×9 racked packageCase of 486$558.90Each$558.90Supplier Number: 609103
Insert caps
Informations sur le produit
- Catalog #DésignationColorAvailabilityPrice