Cubis® II Balances
Cubis® II Balances
Supplier:   Sartorius
Cubis® II Balances
Supplier:  Sartorius
Certificates
Cubis® II Balances
8 Options Available Below
Product Details & Documents
The modular weighing system Cubis® II can be individually configured from different components. The combinability of the display unit, weighing module, draft shield, software packages for various applications and functions and a comprehensive range of accessories allows the individual adaptation of the Cubis® II balance to all weighing tasks.
  • Leading performance
  • Monolithical weighing system, integrated climate sensors, individual sample holders
  • Error-free operation
  • Individual QApp workflows
  • Motorized auto-leveling
  • Full end-to-end data integrity
  • Integrated audit-trail, state-of-the-art user management
  • Outstanding service support
  • Integrated status center, integrated service functions preventive maintenance on accredited standards
The Cubis® II range of premium laboratory balances with a maximum load between 2.1 g and 70 kg and a readability between 0.1 μg to 1 g provide the ideal model for every application.
Connection type: D-sub connector, 9-pin
Display type: Color touchscreen
Certifications: 21 CFR Part 11 compliance.
Specifications for Product Family
Spécifications
  • Catalog No:
  • CA77547-558
  • CA77591-718
  • CA77591-720
  • CA77591-722
  • CA77591-724
  • CA77591-726
  • CA77547-560
  • CA77547-562
  • Modèle:
  • MCA125S
  • MCA125S
  • MCA225S
  • MCA226S
  • MCE125S
  • MCE225S
  • MCA8202S
  • MCE8202S
  • Operating temperature range:
  • 0…40 °C
  • 0…40 °C
  • 0…40 °C
  • 0…40 °C
  • 0…40 °C
  • 0…40 °C
  • 0…40 °C
  • 0…40 °C
  • Plug type:
  • Two-sided plug
  • Two-sided plug
  • Two-sided plug
  • Two-sided plug
  • Two-sided plug
  • Two-sided plug
  • Two-sided plug
  • Two-sided plug
  • Communication interface:
  • COM-RS232 Interface
  • COM-RS232 Interface
  • COM-RS232 Interface
  • COM-RS232 Interface
  • COM-RS232 Interface
  • COM-RS232 Interface
  • COM-RS232 Interface
  • COM-RS232 Interface
  • Voltage:
  • 0 - 240 VAC (±10%)
  • 0 - 240 VAC (±10%)
  • 0 - 240 VAC (±10%)
  • 0 - 240 VAC (±10%)
  • 0 - 240 VAC (±10%)
  • 0 - 240 VAC (±10%)
  • 0 - 240 VAC (±10%)
  • 0 - 240 VAC (±10%)
Spécifications
Product Family Options
Models with U Type draft shield
Informations sur le produit
  • Désignation
    Modèle
    Weighing Capacity
    Linearity
    Plateau de pesée/taille de la plateforme
    Readability
    Stabilization Time
    Repeatabilty
    Dimensions
    Availability
    Price
  • Cubis II Semi-micro Balance, Scale div, (d) 0.01 mg, max. load 120 g. Draft shield U
    MCA125S
    120 g
    0.065 mg
    85×85 mm
    0.01 mg
    2 s
    0.01 mg
    355×240×61 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 19 015,68 $
    19 015,68 $
    Each
       19 015,68 $
    Minimum order quantity is 1
    Supplier Number: MCA125S-U-MTS
    Spécifications:
    More
    Specifications
    Cat. No.
    CA77547-558
    Modèle
    MCA125S
    Operating temperature range
    0…40 °C
    Plug type
    Two-sided plug
    Communication interface
    COM-RS232 Interface
    Voltage
    0 - 240 VAC (±10%)
Models with D Type draft shield
Informations sur le produit
  • Désignation
    Modèle
    Weighing Capacity
    Linearity
    Plateau de pesée/taille de la plateforme
    Readability
    Stabilization Time
    Repeatabilty
    Dimensions
    Availability
    Price
  • Cubis II Semi-micro Balance, Scale div, (d) 0.01 mg, max. load 120 g. Draft shield D
    MCA125S
    120 g
    0.03 mg
    85×85 mm
    0.01 mg
    1.5 s
    0.01 mg
    355×240×61 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 27 479,17 $
    27 479,17 $
    Each
       27 479,17 $
    Minimum order quantity is 1
    Supplier Number: MCA125S-I-3-MTS
    Spécifications:
    More
    Specifications
    Cat. No.
    CA77591-718
    Modèle
    MCA125S
    Operating temperature range
    0…40 °C
    Plug type
    Two-sided plug
    Communication interface
    COM-RS232 Interface
    Voltage
    0 - 240 VAC (±10%)
  • Cubis II Semi-micro Balance, Scale div, (d) 0.01 mg, max. load 220 g. Draft shield D
    MCA225S
    220 g
    0.03 mg
    85×85 mm
    0.01 mg
    1.5 s
    0.01 mg
    355×240×61 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 35 619,76 $
    35 619,76 $
    Each
       35 619,76 $
    Minimum order quantity is 1
    Supplier Number: MCA225S-I-3-MTS
    Spécifications:
    More
    Specifications
    Cat. No.
    CA77591-720
    Modèle
    MCA225S
    Operating temperature range
    0…40 °C
    Plug type
    Two-sided plug
    Communication interface
    COM-RS232 Interface
    Voltage
    0 - 240 VAC (±10%)
  • Cubis II Semi-micro Balance, Scale div, (d) 0.05 mg, max. load 220 g. Draft shield D
    MCA226S
    220 g
    0.03 mg
    85×85 mm
    0.01 mg
    1.5 s
    0.01 mg
    355×240×61 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 40 734,83 $
    40 734,83 $
    Each
       40 734,83 $
    Minimum order quantity is 1
    Supplier Number: MCA226S-I-3-MTS
    Spécifications:
    More
    Specifications
    Cat. No.
    CA77591-722
    Modèle
    MCA226S
    Operating temperature range
    0…40 °C
    Plug type
    Two-sided plug
    Communication interface
    COM-RS232 Interface
    Voltage
    0 - 240 VAC (±10%)
  • Cubis II Semi-micro Balance, Scale div, (d) 0.01 mg, max. load 120 g. Draft shield D
    MCE125S
    120 g
    0.03 mg
    85×85 mm
    0.01 mg
    1.5 s
    0.01 mg
    315×240×61 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 22 775,09 $
    22 775,09 $
    Each
       22 775,09 $
    Minimum order quantity is 1
    Supplier Number: MCE125S-I-3-MTS
    Spécifications:
    More
    Specifications
    Cat. No.
    CA77591-724
    Modèle
    MCE125S
    Operating temperature range
    0…40 °C
    Plug type
    Two-sided plug
    Communication interface
    COM-RS232 Interface
    Voltage
    0 - 240 VAC (±10%)
  • Cubis II Semi-micro Balance, Scale div, (d) 0.01 mg, max. load 220 g. Draft shield D
    MCE225S
    220 g
    0.03 mg
    85×85 mm
    0.01 mg
    1.5 s
    0.01 mg
    315×240×61 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 30 915,66 $
    30 915,66 $
    Each
       30 915,66 $
    Minimum order quantity is 1
    Supplier Number: MCE225S-I-3-MTS
    Spécifications:
    More
    Specifications
    Cat. No.
    CA77591-726
    Modèle
    MCE225S
    Operating temperature range
    0…40 °C
    Plug type
    Two-sided plug
    Communication interface
    COM-RS232 Interface
    Voltage
    0 - 240 VAC (±10%)
Models without draft shield
Informations sur le produit
  • Désignation
    Modèle
    Weighing Capacity
    Linearity
    Plateau de pesée/taille de la plateforme
    Readability
    Stabilization Time
    Repeatabilty
    Dimensions
    Availability
    Price
  • Cubis II Precision Balance, Scale div, (d) 10 mg, max. load 8200 g
    MCA8202S
    8200 g
    6 mg
    206×206 mm
    10 mg
    1 s
    4 mg
    425×240×95 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 13 604,19 $
    13 604,19 $
    Each
       13 604,19 $
    Minimum order quantity is 1
    Supplier Number: MCA8202S-0-MTS
    Spécifications:
    More
    Specifications
    Cat. No.
    CA77547-560
    Modèle
    MCA8202S
    Operating temperature range
    0…40 °C
    Plug type
    Two-sided plug
    Communication interface
    COM-RS232 Interface
    Voltage
    0 - 240 VAC (±10%)
  • Cubis II Precision Balance, Scale div, (d) 10 mg, max. load 8200 g
    MCE8202S
    8200 g
    6 mg
    206×206 mm
    10 mg
    1 s
    4 mg
    425×240×95 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 9 367,00 $
    9 367,00 $
    Each
       9 367,00 $
    Minimum order quantity is 1
    Supplier Number: MCE8202S-0-MTS
    Spécifications:
    More
    Specifications
    Cat. No.
    CA77547-562
    Modèle
    MCE8202S
    Operating temperature range
    0…40 °C
    Plug type
    Two-sided plug
    Communication interface
    COM-RS232 Interface
    Voltage
    0 - 240 VAC (±10%)