3D Barcode Cryogenic Vials
Supplier:   NEST SCIENTIFIC
3D Barcode Cryogenic Vials
Supplier:  NEST SCIENTIFIC
Certificates
3D Barcode Cryogenic Vials
9 Options Available Below
Product Details & Documents
3D Barcode cryogenic vial meets the SBS standard and provides an effective safety guarantee and management for biological sample libraries and other warehouses of various chemicals and biological products stored at low temperatures.
  • Compatible with Brooks system
  • DNase/RNase free, non-endotoxin
  • Electron beam sterilization, SAL = 10⁻⁶
The two-color vial body of an integrated injection molding can realize multi-code tracking, and it can support the traceability of samples and data sharing among multiple users, laboratories and automation, making the warehousing and ex-warehousing of samples more convenient and faster, while ensuring the safety of the sample storage process.
The vial body is made of medical-grade cryogenic polypropylene (PP), which meets USP Class-6 standards, and the vial is recommended to beused in –196 ℃ gas nitrogen phase.
3D Barcode cryogenic vials, 1.8 ml, external thread, double secure cap:
Excellent sealing performance: permanent reversal prevention.
High storage safety: the inside forms a closed and isolated space separated from the outside without leakage, favorable for storage and transportation.
Memory of use history: the middle cover will be punctured when taking sample out, leaving a mark to know it’s been used. This function is to ensure the original fidelity of the sample.
Convenient sampling without contamination: the soft rubber sealing layer can be punctured with a pipette tip or a syringe needle. The outer cover won’t be contaminated by the sample, ensuring safety of the personnel and environment.
Product Family Options
3D Barcode cryogenic vials, SBS format
Informations sur le produit
  • Désignation
    Capacité
    Matière
    Avec bouchon
    Cap Type
    Cap Material
    Sterility
    Packaging
    Availability
    Price
  • 3D Barcode cryogenic vial, SBS format, self-standing, external thread, caps on
    1.9 ml
    PP
    Oui
    Screw cap
    HDPE
    Sterile
    48 vials per pack, 480 per case
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 480
    Case of 480 - $527.61
    $527.61
    Log in pour la disponibilité
    Each
    Each - $527.61
    $527.61
    Case of 480
       $527.61
    Each
       $527.61
    Supplier Number: 612041
  • 3D Barcode cryogenic vial, SBS format, self-standing, external thread, caps separated
    1.9 ml
    PP
    Non
    Screw cap
    HDPE
    Sterile
    48 vials per pack, 480 per case
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 480
    Case of 480 - $470.95
    $470.95
    Log in pour la disponibilité
    Each
    Each - $470.95
    $470.95
    Case of 480
       $470.95
    Each
       $470.95
    Supplier Number: 612047
  • 3D Barcode cryogenic vial, SBS format, self-standing, external thread, racked
    1.9 ml
    PP
    Non
    Screw cap
    HDPE
    Sterile
    48 vials per pack, 480 per case
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 480
    Case of 480 - $577.03
    $577.03
    Log in pour la disponibilité
    Each
    Each - $577.03
    $577.03
    Case of 480
       $577.03
    Each
       $577.03
    Supplier Number: 612051
  • 3D Barcode cryogenic vial, SBS format, self-standing, external thread, caps on
    4 ml
    PP
    Oui
    Screw cap
    HDPE
    Sterile
    48 vials per pack, 480 per case
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 480
    Case of 480 - $572.11
    $572.11
    Log in pour la disponibilité
    Each
    Each - $572.11
    $572.11
    Case of 480
       $572.11
    Each
       $572.11
    Supplier Number: 614041
  • 3D Barcode cryogenic vial, SBS format, self-standing, external thread, caps separated
    4 ml
    PP
    Non
    Screw cap
    HDPE
    Sterile
    48 vials per pack, 480 per case
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 480
    Case of 480 - $531.46
    $531.46
    Log in pour la disponibilité
    Each
    Each - $531.46
    $531.46
    Case of 480
       $531.46
    Each
       $531.46
    Supplier Number: 614047
  • 3D Barcode cryogenic vial, SBS format, self-standing, external thread, racked
    4 ml
    PP
    Non
    Screw cap
    HDPE
    Sterile
    48 vials per pack, 480 per case
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 480
    Case of 480 - $632.63
    $632.63
    Log in pour la disponibilité
    Each
    Each - $632.63
    $632.63
    Case of 480
       $632.63
    Each
       $632.63
    Supplier Number: 614051
3D Barcode cryogenic vials, external thread, double secure cap
Informations sur le produit
  • Désignation
    Capacité
    Matière
    Avec bouchon
    Cap Type
    Cap Material
    Sterility
    Packaging
    Availability
    Price
  • Cryogenic vials, bulk, external thread
    1.8 ml
    PP
    Oui
    Screw cap
    HDPE
    Sterile
    50 vials per pack, 2000 per case
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 2,000
    Case of 2,000 - $1,821.39
    $1,821.39
    Log in pour la disponibilité
    Each
    Each - $1,821.39
    $1,821.39
    Case of 2,000
       $1,821.39
    Each
       $1,821.39
    Supplier Number: 627001
  • Cryogenic vials, 10×10 racked, external thread
    1.8 ml
    PP
    Oui
    Screw cap
    HDPE
    Sterile
    100 vials per pack, 1000 per case
    Discontinued
    Supplier Number: 627002
  • Cryogenic vials, bulk, external thread, double secure cap
    1.8 ml
    PP
    Oui
    Screw cap
    HDPE
    Sterile
    50 vials per pack, 2000 per case
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 2,000
    Case of 2,000 - $2,249.32
    $2,249.32
    Log in pour la disponibilité
    Each
    Each - $2,249.32
    $2,249.32
    Case of 2,000
       $2,249.32
    Each
       $2,249.32
    Supplier Number: 627101
  • Cryogenic vials, 10×10 racked, external thread, double secure cap
    1.8 ml
    PP
    Oui
    Screw cap
    HDPE
    Sterile
    100 vials per pack, 1000 per case
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $1,394.65
    $1,394.65
    Log in pour la disponibilité
    Each
    Each - $1,394.65
    $1,394.65
    Case of 1,000
       $1,394.65
    Each
       $1,394.65
    Supplier Number: 627102