Cell Strainers | Avantor
Cell Strainers
Supplier:   NEST SCIENTIFIC
Cell Strainers
Supplier:  NEST SCIENTIFIC
Certificates
3 Options Available Below
Product Details & Documents
NEST Scientific cell strainers have a PP frame with a nylon strainer. NEST Scientific cell strainers can be used to obtain uniform single cell suspensions and they are compatible with 50 ml centrifuge tubes from most popular brands on the market for ease of use. NEST Scientific cell strainers are ideal for sample filtration, sample preparation for flow cytometry and isolating primary cells to consistently obtain a uniform single cell suspension from tissues.
  • Equipped with a PP frame and nylon strainer
  • Non-pyrogenic, DNase/RNase free and non-cytotoxic
  • Isolating primary cells to consistently obtain a uniform single cell suspension from tissues
  • Sample preparation for flow cytometry
NEST Scientific cell strainers are sterile and individually packaged to maintain sterility of unused units. NEST Scientific cell strainers are non-pyrogenic, DNase and RNase free and non-cytotoxic. They are available in the following filtration specifications or mesh pore size: 40, 70 and 100 µm.
Product Family Options
Informations sur le produit
  • Désignation
    Taille de pore
    Sterility
    Color
    Matière
    Packaging
    Availability
    Price
  • Cell strainer, universal, individually wrapped
    40 µm
    Sterile
    Blue
    PP
    50/case
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $214.21
    $214.21
    Log in pour la disponibilité
    Each
    Each - $214.21
    $214.21
    Case of 50
       $214.21
    Each
       $214.21
    Supplier Number: 258369
  • Cell strainer, universal, individually wrapped
    70 µm
    Sterile
    White
    PP
    50/case
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $216.16
    $216.16
    Log in pour la disponibilité
    Each
    Each - $216.16
    $216.16
    Case of 50
       $216.16
    Each
       $216.16
    Supplier Number: 258368
  • Cell strainer, universal, individually wrapped
    100 µm
    Sterile
    Yellow
    PP
    50/case
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $222.46
    $222.46
    Log in pour la disponibilité
    Each
    Each - $222.46
    $222.46
    Case of 50
       $222.46
    Each
       $222.46
    Supplier Number: 258367