Glassware Carts, Labconco® | Avantor
Glassware Carts, Labconco®
Supplier:   Labconco
Glassware Carts, Labconco®
Supplier:  Labconco
Certificates
Glassware Carts, Labconco®
Glassware Carts, Labconco®
2 Options Available Below
Product Details & Documents
Designed for use with glassware and small equipment
Carts have a welded tubular steel frame with tough, corrosion-resistant glacier white epoxy coating, four 10.2cm (4") diameter casters (two with toe locks), two handles with ergonomic extruded rubber grips, and a heavy-duty removable plastic drip pan. Interchangeable wire baskets are vinyl-coated for glassware safety and easy storage.
Ordering information: Manufacturer's lifetime warranty.
Product Family Options
Informations sur le produit
  • Désignation
    Basket Dimensions
    Weight
    Availability
    Price
  • Cart with 2 Baskets
    32×18×4"
    43 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,520.00
    $1,520.00
    Each
       $1,520.00
    La quantité minimale de commande est de 1
    Supplier Number: 8032500
  • Cart with 4 Baskets
    16×18×4"
    43 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,520.00
    $1,520.00
    Each
       $1,520.00
    La quantité minimale de commande est de 1
    Supplier Number: 8045000
Accessories
Informations sur le produit
  • Catalog #
    Length
    Height
    Availability
    Price
  • Large Replacement Basket, Vinyl Coated Wire, for Cart 82006-868
    Large Replacement Basket, Vinyl Coated Wire, for Cart 82006-868
    19761-050
    81.3 cm (32")
    10.2 cm (4")
    Log in pour la disponibilité
    Each - $370.00
Informations sur le produit
  • Catalog #
    Basket Dimensions
    Length
    Availability
    Price
  • Small Replacement Basket, Vinyl Coated Wire, for Cart 82006-870
    Small Replacement Basket, Vinyl Coated Wire, for Cart 82006-870
    19761-052
    16×18×4"
    40.6 cm (16")
    Log in pour la disponibilité
    Each - $247.00