Whatman™ pH Indicator Papers, on Reels, Whatman products (Cytiva)
Supplier: Cytiva
Certificates
About this item
Can be used for the rapid determination of pH values and has led to many applications in laboratories and industry
- Instant pH readings
- Accurate for a wide range of routine pH testing
- Inexpensive
- Convenient and portable for field use
Three colors roll: The paper has three separate indicator dye colour bands. The individual combination of colour change resulting from each test is compared with the colour-coded comparison chart printed on the dispenser, giving improved speed and accuracy in reading.
Available as standard or three colour ranges as rolls in a handy dispenser.
Frequently Bought Together