Bransonic® Ultrasonic Pipet Cleaner, Branson
CA21812-750
- Capacity:10.4L (2.75 gal.)
- Description:Ultrasonic Pipet Cleaner
- Electrical:115V, 50/60 Hz
- W×D×H:52.1 x 16.5 x 15.2 cm (20.5 x 6.5 x 6")
- Tank W×D×H:50.2x14.6x15.2 cm (19.75 x 5.75 x 6")
- Cat. no.:CA21812-750
- Supplier No.:951-030
For cleaning sampling pipets ultrasonically
Lipids and proteins are removed from the capillary bore of the pipet through the cavitation process.
: Stainless steel cover and pipet basket sold separately.