VWR® Wire Carts with Round Posts
VWR® Wire Carts with Round Posts
Supplier:   VWR International
VWR® Wire Carts with Round Posts
Supplier:  VWR International
Certificates
VWR® Wire Carts with Round Posts
VWR® Wire Carts with Round Posts
48 Options Available Below
Product Details & Documents
Feature bright chrome-plated wire shelves and corner posts.
  • Chrome-plated construction
  • Round post design
  • Adjustable in increments
  • Exceptionally durable
Shelves have truss-reinforced edges for exceptional durability. Shelves are adjustable in 5.1 cm (2") increments. Carts can support up to 544kg (1200lbs.) when weight is evenly distributed.
Ordering information: Supplied with four shelves, four corner posts, and four 12.7 cm (5") swivel casters.
Product Family Options
Product Information
  • Load Capacity
    Largeur
    Hauteur
    Profondeur
    Shipping Weight
    Availability
    Price
  • 1200 lbs.
    36"
    60"
    18"
    62 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $790.64
    $790.64
    Each
       $790.64
  • 1200 lbs.
    48"
    60"
    18"
    72 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $866.00
    $866.00
    Each
       $866.00
  • 1200 lbs.
    60"
    60"
    18"
    92 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $936.31
    $936.31
    Each
       $936.31
  • 1200 lbs.
    72"
    60"
    18"
    104 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $1,039.95
    $1,039.95
    Each
       $1,039.95
  • 1200 lbs.
    36"
    60"
    24"
    76 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $882.31
    $882.31
    Each
       $882.31
  • 1200 lbs.
    48"
    60"
    24"
    88 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $966.21
    $966.21
    Each
       $966.21
  • 1200 lbs.
    60"
    60"
    24"
    108 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $1,061.98
    $1,061.98
    Each
       $1,061.98
  • 1200 lbs.
    72"
    60"
    24"
    128 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $1,177.58
    $1,177.58
    Each
       $1,177.58
  • 1200 lbs.
    36"
    68"
    18"
    64 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $792.92
    $792.92
    Each
       $792.92
  • 1200 lbs.
    48"
    68"
    18"
    64 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $873.24
    $873.24
    Each
       $873.24
  • 1200 lbs.
    60"
    68"
    18"
    94 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $939.35
    $939.35
    Each
       $939.35
  • 1200 lbs.
    72"
    68"
    18"
    106 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $1,046.04
    $1,046.04
    Each
       $1,046.04
  • 1200 lbs.
    36"
    68"
    24"
    78 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $888.38
    $888.38
    Each
       $888.38
  • 1200 lbs.
    48"
    68"
    24"
    90 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $971.45
    $971.45
    Each
       $971.45
  • 1200 lbs.
    60"
    68"
    24"
    110 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $1,075.25
    $1,075.25
    Each
       $1,075.25
  • 1200 lbs.
    72"
    68"
    24"
    130 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $1,189.28
    $1,189.28
    Each
       $1,189.28
  • 1200 lbs.
    36"
    80"
    18"
    66 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $818.56
    $818.56
    Each
       $818.56
  • 1200 lbs.
    48"
    80"
    18"
    71 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $879.47
    $879.47
    Each
       $879.47
  • 1200 lbs.
    60"
    80"
    18"
    112 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $957.22
    $957.22
    Each
       $957.22
  • 1200 lbs.
    72"
    80"
    18"
    118 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $1,047.04
    $1,047.04
    Each
       $1,047.04
  • 1200 lbs.
    36"
    80"
    24"
    84 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $899.17
    $899.17
    Each
       $899.17
  • 1200 lbs.
    48"
    80"
    24"
    92 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $994.03
    $994.03
    Each
       $994.03
  • 1200 lbs.
    60"
    80"
    24"
    112 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $1,098.48
    $1,098.48
    Each
       $1,098.48
  • 1200 lbs.
    72"
    80"
    24"
    124 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $1,198.48
    $1,198.48
    Each
       $1,198.48
  • 1200 lbs.
    36"
    60"
    18"
    62 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $1,514.22
    $1,514.22
    Each
       $1,514.22
  • 1200 lbs.
    48"
    60"
    18"
    72 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $1,653.69
    $1,653.69
    Each
       $1,653.69
  • 1200 lbs.
    60"
    60"
    18"
    92 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $1,803.38
    $1,803.38
    Each
       $1,803.38
  • 1200 lbs.
    72"
    60"
    18"
    104 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $1,979.17
    $1,979.17
    Each
       $1,979.17
  • 1200 lbs.
    36"
    60"
    24"
    76 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $1,679.56
    $1,679.56
    Each
       $1,679.56
  • 1200 lbs.
    48"
    60"
    24"
    88 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $1,865.56
    $1,865.56
    Each
       $1,865.56
  • 1200 lbs.
    60"
    60"
    24"
    108 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $2,056.88
    $2,056.88
    Each
       $2,056.88
  • 1200 lbs.
    72"
    60"
    24"
    128 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $2,273.81
    $2,273.81
    Each
       $2,273.81
  • 1200 lbs.
    36"
    68"
    18"
    64 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $1,524.54
    $1,524.54
    Each
       $1,524.54
  • 1200 lbs.
    48"
    68"
    18"
    64 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $1,664.03
    $1,664.03
    Each
       $1,664.03
  • 1200 lbs.
    60"
    68"
    18"
    94 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $1,813.98
    $1,813.98
    Each
       $1,813.98
  • 1200 lbs.
    72"
    68"
    18"
    106 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $1,989.66
    $1,989.66
    Each
       $1,989.66
  • 1200 lbs.
    36"
    68"
    24"
    78 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $1,689.82
    $1,689.82
    Each
       $1,689.82
  • 1200 lbs.
    48"
    68"
    24"
    90 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $1,875.89
    $1,875.89
    Each
       $1,875.89
  • 1200 lbs.
    60"
    68"
    24"
    110 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $2,067.10
    $2,067.10
    Each
       $2,067.10
  • 1200 lbs.
    72"
    68"
    24"
    130 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $2,284.06
    $2,284.06
    Each
       $2,284.06
  • 1200 lbs.
    36"
    80"
    18"
    66 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $1,555.42
    $1,555.42
    Each
       $1,555.42
  • 1200 lbs.
    48"
    80"
    18"
    71 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $1,695.13
    $1,695.13
    Each
       $1,695.13
  • 1200 lbs.
    60"
    80"
    18"
    112 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $1,844.88
    $1,844.88
    Each
       $1,844.88
  • 1200 lbs.
    72"
    80"
    18"
    118 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $2,020.54
    $2,020.54
    Each
       $2,020.54
  • 1200 lbs.
    36"
    80"
    24"
    84 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $1,720.84
    $1,720.84
    Each
       $1,720.84
  • 1200 lbs.
    48"
    80"
    24"
    92 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $1,906.94
    $1,906.94
    Each
       $1,906.94
  • 1200 lbs.
    60"
    80"
    24"
    112 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $2,098.16
    $2,098.16
    Each
       $2,098.16
  • 1200 lbs.
    72"
    80"
    24"
    124 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $2,315.26
    $2,315.26
    Each
       $2,315.26
Accessories
Product Information
  • Catalog #
    Material
    Color
    Availability
    Price
  • Horizontal triple glove box holder
    Horizontal triple glove box holder
    76423-256
    Epoxy
    Grey
    Log in for availability
    Each - $97.30
Product Information
  • Catalog #
    Color
    For Use With
    Availability
    Price
  • Sanitizer holder, 1 gal., Metroseal gray epoxy
    Sanitizer holder, 1 gal., Metroseal gray epoxy
    76423-248
    Grey
    SmartWall and Super Erecta shelving
    Log in for availability
    Each - $78.40
Product Information
  • Catalog #
    Material
    Color
    Availability
    Price
  • Horizontal single glove box holder
    Horizontal single glove box holder
    76423-254
    Epoxy
    Grey
    Log in for availability
    Each - $78.40
Product Information
  • Catalog #
    Color
    For Use With
    Availability
    Price
  • Sanitizer holder, 2 L, Metroseal gray epoxy
    Sanitizer holder, 2 L, Metroseal gray epoxy
    76423-250
    Grey
    SmartWall and Super Erecta shelving
    Log in for availability
    Each - $72.10