PYREX® Polyethylene [ST] Stoppers, Corning
Supplier: Corning
PYREX® Polyethylene [ST] Stoppers, Corning
Supplier: Corning
Certificates
À propos de cet article
These PYREX® No.13 stoppers are made from linear HDPE and have closed bottoms to conform to Standard Taper stopper dimensions.
- Blue and white with a closed bottom
- Three rings on the plug provide an efficient seal
- Improved design fully protects the neck from accidental tipping
9 Options Available Below
Product Details & Documents
These PYREX® No.13 stoppers are made from linear HDPE and have closed bottoms to conform to Standard Taper stopper dimensions.
- Blue and white with a closed bottom
- Three rings on the plug provide an efficient seal
- Improved design fully protects the neck from accidental tipping
Specifications for Product Family
Spécifications
- Catalog No:
- 59492-041
- 59492-085
- 59492-100
- 59492-125
- 59492-187
- 59492-039
- 59492-063
- 59492-121
- 59492-123
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
Spécifications
Product Family Options
Informations sur le produit
- DésignationTailleST Stopper No.AvailabilityPrice
- 9Log in pour la disponibilitéLog in pour la disponibilitéCase of 6Case of 6 - $35.19$35.19Prix promotionnel pour les nouveaux clientsCase of 6$35.19Supplier Number: 7624-9Spécifications:Product available on GSA Advantage®
Low Manufacturing Impact More
Specifications
Cat. No.59492-041Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationI-RECs, ISO 14001 - Monterrey - Supplier Number: 7624-16Spécifications:Product available on GSA Advantage®
Low Manufacturing Impact More
Specifications
Cat. No.59492-085Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationI-RECs, ISO 14001 - Monterrey - Supplier Number: 7624-19Spécifications:Product available on GSA Advantage®
Low Manufacturing Impact More
Specifications
Cat. No.59492-100Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationI-RECs, ISO 14001 - Monterrey - Supplier Number: 7624-32Spécifications:Product available on GSA Advantage®
Low Manufacturing Impact More
Specifications
Cat. No.59492-125Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationI-RECs, ISO 14001 - Monterrey - Supplier Number: 7624-38Spécifications:Product available on GSA Advantage®
Low Manufacturing Impact More
Specifications
Cat. No.59492-187Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationI-RECs, ISO 14001 - Monterrey - Supplier Number: 7624-8Spécifications:Product available on GSA Advantage®
Low Manufacturing Impact More
Specifications
Cat. No.59492-039Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationI-RECs, ISO 14001 - Monterrey - 13Log in pour la disponibilitéLog in pour la disponibilitéCase of 6Case of 6 - $38.72$38.72Prix promotionnel pour les nouveaux clientsCase of 6$38.72Supplier Number: 7624-13Spécifications:Product available on GSA Advantage®
Low Manufacturing Impact More
Specifications
Cat. No.59492-063Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationI-RECs, ISO 14001 - Monterrey - 22Log in pour la disponibilitéLog in pour la disponibilitéCase of 6Case of 6 - $58.05$58.05Prix promotionnel pour les nouveaux clientsCase of 6$58.05Supplier Number: 7624-22Spécifications:Product available on GSA Advantage®
Low Manufacturing Impact More
Specifications
Cat. No.59492-121Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationI-RECs, ISO 14001 - Monterrey - Supplier Number: 7624-27Spécifications:Product available on GSA Advantage®
Low Manufacturing Impact More
Specifications
Cat. No.59492-123Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationI-RECs, ISO 14001 - Monterrey