Synthware PTFE Stoppers, Solid and Hollow
Supplier: Kemtech America
Jump to:
About this item
PTFE stopper features a nylon extraction nut that permits easy removal of the stopper from the glass outer joint.
- Extraction nut
- Teflon® construction
Available in solid or hollow formats.
4 Options Available Below
Product Details & Documents
PTFE stopper features a nylon extraction nut that permits easy removal of the stopper from the glass outer joint.
- Extraction nut
- Teflon® construction
Available in solid or hollow formats.