SmartTray™ System, Metro™
SmartTray™ System, Metro™
Supplier:   Metropolitan Wire
SmartTray™ System, Metro™
Supplier:  Metropolitan Wire
Certificates
SmartTray™ System, Metro™
SmartTray™ System, Metro™
7 Options Available Below
Product Details & Documents
Designed to maximize productivity by accelerating throughput and minimizing PCB handling damage
Premium ESD tray inlay (22221-396) offers maximum ESD protection in addition to superior resistance to heat, abrasion, and chemicals. The soft dissipative top layer makes it ideal for use in soldering and assembly areas. Economy tray inlays are not recommended for use in soldering applications.
This cart and tray system facilitates efficient tracking of work-in-process and finished goods with the use of bar code technology. Carts and trays can also be color-coded for easy sorting and categorizing through the use of colored labels (not included). A detent system allows pulled-out trays to be suspended from the cart for optimum ergonomic access. Molded from Metrostat™ material, system provides premium ESD protection for sensitive electronic parts. SmartTray trays can be used with all bottom-load tray cart designs, Metro and non-Metro models, as well as conveyor systems.
Ordering information: Each cart comes complete with four vibration suppression casters (two brake and two swivel), a grounding cable, and two stop bars. Short SmartTray carts include two push handles. Color-coding labels are available; contact your Avantor representative for more information.
Product Family Options
Informations sur le produit
  • Désignation
    Load Capacity
    Longueur
    Largeur
    Hauteur
    Weight
    Availability
    Price
  • Front-Load Cart with 20 SmartTrays and 20 Premium Tray Inlays, Short SmartTray Cart
    25 lbs./level, 500 lbs./cart
    22"
    28"
    49"
    160 lbs. with trays and inlays, 80 lbs. cart only
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $7,195.12
    $7,195.12
    Each
       $7,195.12
    Supplier Number: CBNTC20MSOL1
  • Front-Load Cart with 30 SmartTrays and 30 Premium Tray Inlays, Tall SmartTray Cart
    20 lbs./ level, 600 lbs./cart
    22"
    28"
    63"
    217 lbs. with trays and inlays, 97 lbs. cart only
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $9,665.17
    $9,665.17
    Each
       $9,665.17
    Supplier Number: CBNTC30MSOL1
  • Front-Load Cart with 20 SmartTrays and 20 Economy Tray Inlays, Short SmartTray Cart
    25 lbs./level, 500 lbs./cart
    22"
    28"
    49"
    160 lbs. with trays and inlays, 80 lbs. cart only
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $6,795.00
    $6,795.00
    Each
       $6,795.00
    Supplier Number: CBNTC20MSOL2
  • Front-Load Cart with 30 SmartTrays and 30 Economy Tray Inlays, Tall SmartTray Cart
    20 lbs./ level, 600 lbs./cart
    22"
    28"
    63"
    217 lbs. with trays and inlays, 97 lbs. cart only
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $9,056.06
    $9,056.06
    Each
       $9,056.06
    Supplier Number: CBNTC30MSOL2
  • Front-Load Cart Only, Holds 20 Trays, Short SmartTray Cart
    25 lbs./level, 500 lbs./cart
    22"
    28"
    49"
    160 lbs. with trays and inlays, 80 lbs. cart only
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,654.18
    $2,654.18
    Each
       $2,654.18
    Supplier Number: CBNTC20M
  • Front-Load Cart Only, Holds 30 Trays, Tall SmartTray Cart
    20 lbs./ level, 600 lbs./cart
    22"
    28"
    63"
    217 lbs. with trays and inlays, 97 lbs. cart only
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,919.55
    $2,919.55
    Each
       $2,919.55
    Supplier Number: CBNTC30M
  • Side-Load Cart Only, Holds 20 Trays, Short SmartTray Cart
    25 lbs./level, 500 lbs./cart
    30"
    22"
    49"
    160 lbs. with trays and inlays, 80 lbs. cart only
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,654.18
    $2,654.18
    Each
       $2,654.18
    Supplier Number: CBNTCS20M
Accessories
Informations sur le produit
  • Catalog #
    Length
    Width
    Availability
    Price
  • Premium Rubber ESD Inlay, Two-Layer, Blue Top/Black Bottom
    Premium Rubber ESD Inlay, Two-Layer, Blue Top/Black Bottom
    CA22221-396
    16⅛"
    24⅛"
    Log in pour la disponibilité
    Each - $136.95