Synthware Stopper, Cap Design, Kemtech America
Designed for use with an apparatus equipped with an inner opening.
- Heavy wall borosilicate construction
- Solid glass plug
- Double oblique
- Retention clip lncluded Female Ground glass joint
For apparatus having an inner opening.
Frequently Bought Together