VWR® Corks
Assorted, Nos. 12 to 26
CA23417-000
XXXX Corks function as a closure for bottles, vials, laboratory and other openings.
- Regular length taper
- Assorted sizes
The XXXX premium grade corks have fewer lenticels (crevices) than standard grade. This means they are better suited for use as laboratory corks and for transporting many kinds of liquids.
- Désignation:Assorted, Nos. 12 to 26
- Cat. No.:CA23417-000
- Environmentally Preferable:Low Manufacturing Impact