Process Cooling Box | Avantor
Process Cooling Box
Supplier:   NEST SCIENTIFIC
Process Cooling Box
Supplier:  NEST SCIENTIFIC
Certificates
Process Cooling Box
2 Options Available Below
Product Details & Documents
This product can used in variety of cell types, including stem cells, primary cells, cell lines and yeast. The cooling box does not require any liquid or additive, all you need is placed the cool box into the ultra-low temperature refrigerator, then it can ensure that the box's has the cooling rate of −1 ℃ per minute.
  • It is a very good device for all types cell preservation
  • The product uses a symmetrical design with a combination on of foam and precious alloys
  • No additive and no emissions characteristics
  • It is an environmental friendly products
  • It does not need to replace any parts, and can be permanently used
This product is easy to use, just insert the frozen tube, placed the unit in −80 ℃ freeze environment. It can greatly improve the handling efficiency and has a good survival rate of cell preservation. The sample is safer and worry free.
It holds upto 12 tubes - 1/2 ml cryogenic vials or 1.5/2 ml centrifuge tubes.
Product Family Options
Informations sur le produit
  • Désignation
    Dimensions
    Weight
    Availability
    Price
  • Process cooling box for 12 tubes, Hexagonal
    Side length: 108×100 mm
    120 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1
    Case of 1 - $528.09
    $528.09
    Log in pour la disponibilité
    Each
    Each - $528.09
    $528.09
    Case of 1
       $528.09
    Each
       $528.09
    Supplier Number: 200101
  • Process cooling box for 12 tubes, square
    117×117×100 mm
    160 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1
    Case of 1 - $528.09
    $528.09
    Log in pour la disponibilité
    Each
    Each - $528.09
    $528.09
    Case of 1
       $528.09
    Each
       $528.09
    Supplier Number: 200102