Storage Bottles, Round
Storage Bottles, Round
Storage Bottles, Round
Storage Bottles, Round
Round Storage Bottles are manufactured in 100,000 grade cleaning room with USP 6 grade raw materials.
  • Excellent physical and chemical indicators
  • Good pressure-resistance
  • Impact-resistance
  • Acid and alkali resistance
  • Autoclave sterilization at 121 ℃, HDPE can be stored at −80 ℃
  • No biotoxicity, pyrogen-free, DNAse/RNAse-free
Sterile and enzyme-free plastic reagent bottles, suitable for product packaging and storage requirements in molecular biology and Cell Biology, laboratory medicine, genomics and proteomics and other areas. Improvement of the quality of product packaging helps to improve the company image and brand value, lets your product packaging meet international standards and enter the international market.
Washing-free, no need for tedious pre-cleaning processing work, ready to use, greatly improves the packaging effect of users. Thickened middle packaging to ensure transportation and storage safety. Use of professional leaking-proof design for bottle mouth, with no need for inner cover or inner gasket for protection, excellent sealability. Guarantee of being 100% leaking-proof to ensure safety even during air transportation, wide-mouthed design for easy access of liquid.
Certifications: ISO 9001&13485 certified.
Product Family Options
Amber bottles
  • Matière
    Color
    Volume
    Bottle Style
    Packaging
  • PP
    Amber
    8 ml
    Round
    20/pack, 400/case
    Case of 400
    Case of 400 - $282.52
    $282.52
    Each
    Each - $282.52
    $282.52
    Case of 400
       $282.52
    Each
       $282.52
     : 335201
  • HDPE
    Amber
    8 ml
    Round
    20/pack, 400/case
    Case of 400
    Case of 400 - $278.95
    $278.95
    Each
    Each - $278.95
    $278.95
    Case of 400
       $278.95
    Each
       $278.95
     : 335301
  • PP
    Amber
    15 ml
    Round
    20/pack, 400/case
    Case of 400
    Case of 400 - $319.38
    $319.38
    Each
    Each - $319.38
    $319.38
    Case of 400
       $319.38
    Each
       $319.38
     : 336201
  • HDPE
    Amber
    15 ml
    Round
    20/pack, 400/case
    Case of 400
    Case of 400 - $307.40
    $307.40
    Each
    Each - $307.40
    $307.40
    Case of 400
       $307.40
    Each
       $307.40
     : 336301
  • PP
    Amber
    30 ml
    Round
    10/pack, 200/case
    Case of 200
    Case of 200 - $253.24
    $253.24
    Each
    Each - $253.24
    $253.24
    Case of 200
       $253.24
    Each
       $253.24
     : 337201
  • HDPE
    Amber
    30 ml
    Round
    10/pack, 200/case
    Case of 200
    Case of 200 - $260.00
    $260.00
    Each
    Each - $260.00
    $260.00
    Case of 200
       $260.00
    Each
       $260.00
     : 337301
  • PP
    Amber
    60 ml
    Round
    10/pack, 200/case
    Case of 200
    Case of 200 - $297.87
    $297.87
    Each
    Each - $297.87
    $297.87
    Case of 200
       $297.87
    Each
       $297.87
     : 338201
  • HDPE
    Amber
    60 ml
    Round
    10/pack, 200/case
    Case of 200
    Case of 200 - $305.46
    $305.46
    Each
    Each - $305.46
    $305.46
    Case of 200
       $305.46
    Each
       $305.46
     : 338301
  • PP
    Amber
    125 ml
    Round
    10/pack, 100/case
    Case of 100
    Case of 100 - $249.97
    $249.97
    Each
    Each - $249.97
    $249.97
    Case of 100
       $249.97
    Each
       $249.97
     : 339201
  • HDPE
    Amber
    125 ml
    Round
    10/pack, 100/case
    Case of 100
    Case of 100 - $256.10
    $256.10
    Each
    Each - $256.10
    $256.10
    Case of 100
       $256.10
    Each
       $256.10
     : 339301
  • PP
    Amber
    250 ml
    Round
    10/pack, 100/case
    Case of 100
    Case of 100 - $309.04
    $309.04
    Each
    Each - $309.04
    $309.04
    Case of 100
       $309.04
    Each
       $309.04
     : 340201
  • HDPE
    Amber
    250 ml
    Round
    10/pack, 100/case
    Case of 100
    Case of 100 - $313.96
    $313.96
    Each
    Each - $313.96
    $313.96
    Case of 100
       $313.96
    Each
       $313.96
     : 340301
  • PP
    Amber
    500 ml
    Round
    10/pack, 50/case
    Case of 50
    Case of 50 - $273.91
    $273.91
    Each
    Each - $273.91
    $273.91
    Case of 50
       $273.91
    Each
       $273.91
     : 341201
  • HDPE
    Amber
    500 ml
    Round
    10/pack, 50/case
    Case of 50
    Case of 50 - $277.81
    $277.81
    Each
    Each - $277.81
    $277.81
    Case of 50
       $277.81
    Each
       $277.81
     : 341301
White bottles
  • Matière
    Color
    Volume
    Bottle Style
    Packaging
  • HDPE
    White
    8 ml
    Round
    20/pack, 400/case
    Case of 400
    Case of 400 - $245.28
    $245.28
    Each
    Each - $245.28
    $245.28
    Case of 400
       $245.28
    Each
       $245.28
     : 335101
  • HDPE
    White
    15 ml
    Round
    20/pack, 400/case
    Case of 400
    Case of 400 - $269.06
    $269.06
    Each
    Each - $269.06
    $269.06
    Case of 400
       $269.06
    Each
       $269.06
     : 336101
  • HDPE
    White
    30 ml
    Round
    10/pack, 200/case
    Case of 200
    Case of 200 - $236.78
    $236.78
    Each
    Each - $236.78
    $236.78
    Case of 200
       $236.78
    Each
       $236.78
     : 337101
  • HDPE
    White
    60 ml
    Round
    10/pack, 200/case
    Case of 200
    Case of 200 - $267.60
    $267.60
    Each
    Each - $267.60
    $267.60
    Case of 200
       $267.60
    Each
       $267.60
     : 338101
  • HDPE
    White
    125 ml
    Round
    10/pack, 100/case
    Case of 100
    Case of 100 - $227.32
    $227.32
    Each
    Each - $227.32
    $227.32
    Case of 100
       $227.32
    Each
       $227.32
     : 339101
  • HDPE
    White
    250 ml
    Round
    10/pack, 100/case
    Case of 100
    Case of 100 - $274.72
    $274.72
    Each
    Each - $274.72
    $274.72
    Case of 100
       $274.72
    Each
       $274.72
     : 340101
  • HDPE
    White
    500 ml
    Round
    10/pack, 50/case
    Case of 50
    Case of 50 - $244.31
    $244.31
    Each
    Each - $244.31
    $244.31
    Case of 50
       $244.31
    Each
       $244.31
     : 341101