Nalgene® Rectangular Tanks, Low-Density Polyethylene, Thermo Scientific
Nalgene® Rectangular Tanks, Low-Density Polyethylene, Thermo Scientific
Nalgene® Rectangular Tanks, Low-Density Polyethylene, Thermo Scientific
Nalgene® Rectangular Tanks, Low-Density Polyethylene, Thermo Scientific
Nalgene® Rectangular Tanks, Low-Density Polyethylene, Thermo Scientific
Tanks have stepped rim for strength and overlapping skirted cover
Molded in one piece with good chemical resistance
Ordering information: Tanks can be modified with factory-installed spigots; contact your Avantor sales representative for more information.
Product Family Options
  • Volume
    L×W×D
  • 8 L (2 gal.)
    20 x 20 x 20 cm (8 x 8 x 8")
    Each
    Each - $309.60
    $309.60
    Each
       $309.60
     : 14100-0002
  • 20 L (5 gal.)
    25 x 25 x 36 cm (10 x 10 x 14")
    Each
    Each - $388.00
    $388.00
    Each
       $388.00
     : 14100-0005
  • 30 L (8 gal.)
    30 x 30 x 30 cm (12 x 12 x 12")
    Each
    Each - $390.40
    $390.40
    Each
       $390.40
     : 14100-0010
  • 38 L (10 gal.)
    46 x 30 x 30 cm (18 x 12 x 12")
    Each
    Each - $510.40
    $510.40
    Each
       $510.40
     : 14100-0015
  • 50 L (13 gal.)
    61 x 30 x 30 cm (24 x 12 x 12")
    Each
    Each - $591.20
    $591.20
    Each
       $591.20
     : 14100-0020
  • 60 L (16 gal.)
    45 x 30 x 45 cm (18 x 12 x 18")
    Each
    Each - $627.20
    $627.20
    Each
       $627.20
     : 14100-0040
  • 120 L (32 gal.)
    61 x 45 x 45 cm (24 x 18 x 18")
    Each
    Each - $864.00
    $864.00
    Each
       $864.00
     : 14100-0045
  • 160 L (40 gal.)
    61 x 45 x 60 cm (24 x 18 x 24")
    Each
    Each - $1,168.00
    $1,168.00
    Each
       $1,168.00
     : 14100-0065