KIMBLE® MICROFLEX® Williamson Microscale Kits, DWK Life Sciences | Avantor
KIMBLE® MICROFLEX® Williamson Microscale Kits, DWK Life Sciences
KIMBLE® MICROFLEX® Williamson Microscale Kits, DWK Life Sciences
Certificates
KIMBLE® MICROFLEX® Williamson Microscale Kits, DWK Life Sciences
KIMBLE® MICROFLEX® Williamson Microscale Kits, DWK Life Sciences
6 Options Available Below
Product Details & Documents
These organic chemistry kits feature thermoplastic chaste connections and specially tooled glassware components for great versatility to perform Williamson experiments.
Kits are packed in a 34.3Wx25.4Dx6.4H cm (13¹/₂x10x2¹/₂") rugged polyethylene storage case with die-cut foam insert. Kits easily fit in lab bench drawers.
EKONO-CASE™, a low-cost corrugated cardboard alternative to plastic.
Certifications: Glassware is manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Informations sur les accessoires: Test fabric is useful for testing dyes on a variety of fabrics. Three-foot length of fabric four inches wide features 8 strips of different fibers woven side by side. Filament acetate, bleached cotton, spun polyamide type 66, polyester disperse dyeable, spun polyacrylic, spun silk, spun viscose, and worsted wool.
Specifications for Product Family
Spécifications
  • Catalog No:
  • KT748100-0000
  • KT748110-0000
  • KT748000-0000
  • KT748000-0005
  • KT748100-0005
  • KT748110-0005
  • Description:
  • Improved Standard Kit
  • Intermediate Kit
  • Standard Kit
  • Kit contents:
  • 1 Connecting Adapter
    1 Filter Adapter, Pluro Stopper
    1 Connecting Adapter
    1 Filter Adapter, Pluro Stopper
    1Thermometer Adapter
    1 Centrifuge Tube with Cap, 15 ml
    1 Glass Chromatography Column, 150 mm , With Disc
    1 One Way Stopcock, Polyethylene/Nylon
    1 Connector with Support Rod
    1 Connector Only
    1 Connector Only, Viton*
    1 Distillation Column
    1 Distillation Head, 60 mm
    1 Short Neck Flask, 5 ml
    1 Long Neck Flask, 5 ml
    3 Erlenmeyer Flask, 10 ml
    1 Filter Flask, 25 ml
    1 Hirsch Funnel with Disc
    5 Reaction Tube, 10x100 mm
    1 Stainless Steel Spatula
    1 Polyethylene Syringe
    2 PTFE 1.6 mm Tubing, Fits 20G Needle
    1 Pipet, 1 ml
    1 Stir Bars, 4x12 mm, 2 per Vial
  • 1 Connecting Adapter
    1 Filter Adapter, Pluro Stopper
    1 Connecting Adapter
    1 Filter Adapter, Pluro Stopper
    1Thermometer Adapter
    1 Centrifuge Tube with Cap, 15 ml
    1 Glass Chromatography Column, 150 mm , With Disc
    1 One Way Stopcock, Polyethylene/Nylon
    1 Connector with Support Rod
    1 Connector Only, Viton*
    1 Distillation Column
    1 Distillation Head, 60 mm
    1 Short Neck Flask, 5 ml
    1 Filter Flask, 25 ml
    1 Hirsch Funnel with Disc
    2 Reaction Tube, 10x100 mm
    1 Polyethylene Syringe
    2 PTFE 1.6 mm Tubing, Fits 20G Needle
  • 1 Connecting Adapter
    1 Filter Adapter, Pluro Stopper
    1 Connecting Adapter
    1 Filter Adapter, Pluro Stopper
    1Thermometer Adapter
    1 Centrifuge Tube with Cap, 15 ml
    1 Glass Chromatography Column, 150 mm , With Disc
    1 One Way Stopcock, Polyethylene/Nylon
    1 Connector with Support Rod
    1 Connector Only, Viton*
    1 Distillation Column
    1 Distillation Head, 60 mm
    1 Short Neck Flask, 5 ml
    1 Long Neck Flask, 5 ml
    4 Erlenmeyer Flask, 10 ml
    1 Filter Flask, 25 ml
    1 Hirsch Funnel with Disc
    4 Reaction Tube, 10x100 mm
    1 Stainless Steel Spatula
    1 Polyethylene Syringe
    2 PTFE 1.6 mm Tubing, Fits 20G Needle
Spécifications
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Improved standard kit
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $702.95
    $702.95
    Prix promotionnel pour les nouveaux clients
    Each
       $702.95
    Supplier Number: 748100-0000
    Spécifications:
    More
    Specifications
    Cat. No.
    KT748100-0000
    Description
    Improved Standard Kit
    Kit contents
    1 Connecting Adapter
    1 Filter Adapter, Pluro Stopper
    1 Connecting Adapter
    1 Filter Adapter, Pluro Stopper
    1Thermometer Adapter
    1 Centrifuge Tube with Cap, 15 ml
    1 Glass Chromatography Column, 150 mm , With Disc
    1 One Way Stopcock, Polyethylene/Nylon
    1 Connector with Support Rod
    1 Connector Only
    1 Connector Only, Viton*
    1 Distillation Column
    1 Distillation Head, 60 mm
    1 Short Neck Flask, 5 ml
    1 Long Neck Flask, 5 ml
    3 Erlenmeyer Flask, 10 ml
    1 Filter Flask, 25 ml
    1 Hirsch Funnel with Disc
    5 Reaction Tube, 10x100 mm
    1 Stainless Steel Spatula
    1 Polyethylene Syringe
    2 PTFE 1.6 mm Tubing, Fits 20G Needle
    1 Pipet, 1 ml
    1 Stir Bars, 4x12 mm, 2 per Vial
  • Intermediate kit
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $387.66
    $387.66
    Prix promotionnel pour les nouveaux clients
    Each
       $387.66
    Supplier Number: 748110-0000
    Spécifications:
    More
    Specifications
    Cat. No.
    KT748110-0000
    Description
    Intermediate Kit
    Kit contents
    1 Connecting Adapter
    1 Filter Adapter, Pluro Stopper
    1 Connecting Adapter
    1 Filter Adapter, Pluro Stopper
    1Thermometer Adapter
    1 Centrifuge Tube with Cap, 15 ml
    1 Glass Chromatography Column, 150 mm , With Disc
    1 One Way Stopcock, Polyethylene/Nylon
    1 Connector with Support Rod
    1 Connector Only, Viton*
    1 Distillation Column
    1 Distillation Head, 60 mm
    1 Short Neck Flask, 5 ml
    1 Filter Flask, 25 ml
    1 Hirsch Funnel with Disc
    2 Reaction Tube, 10x100 mm
    1 Polyethylene Syringe
    2 PTFE 1.6 mm Tubing, Fits 20G Needle
  • Standard kit
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $719.81
    $719.81
    Prix promotionnel pour les nouveaux clients
    Each
       $719.81
    Supplier Number: 748000-0000
    Spécifications:
    More
    Specifications
    Cat. No.
    KT748000-0000
    Description
    Standard Kit
    Kit contents
    1 Connecting Adapter
    1 Filter Adapter, Pluro Stopper
    1 Connecting Adapter
    1 Filter Adapter, Pluro Stopper
    1Thermometer Adapter
    1 Centrifuge Tube with Cap, 15 ml
    1 Glass Chromatography Column, 150 mm , With Disc
    1 One Way Stopcock, Polyethylene/Nylon
    1 Connector with Support Rod
    1 Connector Only, Viton*
    1 Distillation Column
    1 Distillation Head, 60 mm
    1 Short Neck Flask, 5 ml
    1 Long Neck Flask, 5 ml
    4 Erlenmeyer Flask, 10 ml
    1 Filter Flask, 25 ml
    1 Hirsch Funnel with Disc
    4 Reaction Tube, 10x100 mm
    1 Stainless Steel Spatula
    1 Polyethylene Syringe
    2 PTFE 1.6 mm Tubing, Fits 20G Needle
  • Standard kit, EKONO-CASE™
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $682.50
    $682.50
    Prix promotionnel pour les nouveaux clients
    Each
       $682.50
    Supplier Number: 748000-0005
    Spécifications:
    More
    Specifications
    Cat. No.
    KT748000-0005
    Description
    Kit contents
  • Improved standard kit, EKONO-CASE™
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $721.63
    $721.63
    Prix promotionnel pour les nouveaux clients
    Each
       $721.63
    Supplier Number: 748100-0005
    Spécifications:
    More
    Specifications
    Cat. No.
    KT748100-0005
    Description
    Kit contents
  • Intermediate kit, EKONO-CASE™
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $340.34
    $340.34
    Prix promotionnel pour les nouveaux clients
    Each
       $340.34
    Supplier Number: 748110-0005
    Spécifications:
    More
    Specifications
    Cat. No.
    KT748110-0005
    Description
    Kit contents
Accessories
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Wilfilter centrifuge adapter, Hר: 34×14 mm
    Wilfilter centrifuge adapter, Hר: 34×14 mm
    KT748025-1034
    Log in pour la disponibilité
    Case of 5 - $55.67
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Thermometer adapter
    Thermometer adapter
    KT748003-0000
    Log in pour la disponibilité
    Pack of 10 - $111.02
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Thermometer adapter, accommodation range 5 - 6 mm
    Thermometer adapter, accommodation range 5 - 6 mm
    KT748003-1000
    Log in pour la disponibilité
    Case of 5 - $55.51
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Long neck flask, 5 ml
    Long neck flask, 5 ml
    KT748009-0005
    Log in pour la disponibilité
    Pack of 10 - $470.90
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Filter flask, 25 ml
    Filter flask, 25 ml
    KT748010-1025
    Log in pour la disponibilité
    Each - $64.97
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Filter flask, 25 ml
    Filter flask, 25 ml
    KT748010-0025
    Log in pour la disponibilité
    Pack of 10 - $555.90
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Pipette, 1 ml below bulb
    Pipette, 1 ml below bulb
    KT764100-1001
    Log in pour la disponibilité
    Pack of 25 - $36.58
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Erlenmeyer flask, 10 ml
    Erlenmeyer flask, 10 ml
    KT882300-0010
    Log in pour la disponibilité
    Pack of 12 - $247.52
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Liebig condenser with Williamson Chaste connections, borosilicate glass, height 97 mm
    Liebig condenser with Williamson Chaste connections, borosilicate glass, height 97 mm
    KT748020-1000
    Log in pour la disponibilité
    Each - $153.85
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Connector with support rod
    Connector with support rod
    KT748005-0000
    Log in pour la disponibilité
    Pack of 10 - $292.40
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Connector with support rod
    Connector with support rod
    KT748005-1000
    Log in pour la disponibilité
    Case of 5 - $162.89
Informations sur le produit
  • Catalog #
    Availability
    Price
  • H-connector, thermoplastic
    H-connector, thermoplastic
    KT748005-1001
    Log in pour la disponibilité
    Case of 5 - $67.07
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Connector, FKM
    Connector, FKM
    KT748005-1651
    Log in pour la disponibilité
    Pack of 5 - $89.25
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Connector thermoplastic elastomer
    Connector thermoplastic elastomer
    KT748004-0000
    Log in pour la disponibilité
    Pack of 10 - $135.59
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Connector thermoplastic elastomer
    Connector thermoplastic elastomer
    KT748004-1000
    Log in pour la disponibilité
    Case of 5 - $74.44
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Storage case, 2¹/₂×13¹/₂×10", for Improved standard kit
    Storage case, 2¹/₂×13¹/₂×10", for Improved standard kit
    KT746001-0001
    Log in pour la disponibilité
    Case of 1 - $82.17
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Foam insert set, for Improved standard kit
    Foam insert set, for Improved standard kit
    KT748001-0003
    Log in pour la disponibilité
    Each - $33.49
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Connecting adapter
    Connecting adapter
    KT748001-1000
    Log in pour la disponibilité
    Case of 1 - $58.79
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Filter adapter, pluro stopper
    Filter adapter, pluro stopper
    KT748002-0000
    Log in pour la disponibilité
    Pack of 10 - $48.19
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Short neck flask, 5 ml
    Short neck flask, 5 ml
    KT748008-1005
    Log in pour la disponibilité
    Case of 1 - $54.05
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Short neck flask, 5 ml
    Short neck flask, 5 ml
    KT748008-0005
    Log in pour la disponibilité
    Pack of 10 - $491.40
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Reaction tube, 10×100 mm
    Reaction tube, 10×100 mm
    KT748013-0000
    Log in pour la disponibilité
    Pack of 100 - $743.75
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Reaction tube, 10×100 mm
    Reaction tube, 10×100 mm
    KT748013-1000
    Log in pour la disponibilité
    Case of 25 - $201.45
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Distillation head, 60 mm
    Distillation head, 60 mm
    KT748007-1060
    Log in pour la disponibilité
    Each - $53.21
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Distillation column
    Distillation column
    KT748006-0100
    Log in pour la disponibilité
    Pack of 100 - $1,137.50
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Distillation column
    Distillation column
    KT748006-1100
    Log in pour la disponibilité
    Case of 5 - $60.61
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Polyethylene disc, 20 Microns
    Polyethylene disc, 20 Microns
    KT748017-0020
    Log in pour la disponibilité
    Pack of 100 - $103.70
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Hirsch funnel, with disc
    Hirsch funnel, with disc
    KT748012-0000
    Log in pour la disponibilité
    Pack of 10 - $72.08
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Hirsch funnel, with disc
    Hirsch funnel, with disc
    KT748012-1000
    Log in pour la disponibilité
    Pack of 5 - $39.35
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Multi-fiber test Fabric, 4 W×36" L
    Multi-fiber test Fabric, 4 W×36" L
    10024-800
    Log in pour la disponibilité
    Case of 1 - $22.27
Informations sur le produit
  • Catalog #
    Size
    Availability
    Price
  • Micro spatula, Stainless steel 14/20 or 14/10 outer joint, blade width: 8 mm (0.315"), length: 193.6 mm (7.625")
    Micro spatula, Stainless steel 14/20 or 14/10 outer joint, blade width: 8 mm (0.315"), length: 193.6 mm (7.625")
    KT748014-0000
    Flat-rounded end/v-shaped end
    Log in pour la disponibilité
    Pack of 10 - $353.60
Informations sur le produit
  • Catalog #
    Size
    Availability
    Price
  • Micro spatula, Stainless steel 14/20 or 14/10 outer joint, blade width: 8 mm (0.315"), length: 193.6 mm (7.625")
    Micro spatula, Stainless steel 14/20 or 14/10 outer joint, blade width: 8 mm (0.315"), length: 193.6 mm (7.625")
    KT748014-1000
    Flat-rounded end/v-shaped end
    Log in pour la disponibilité
    Case of 1 - $41.77