Spécifications
- Description:ABS tape dispenser
- Cat. no.:36440-107
- W×D×H:81/8×45/8×23/8"
- Supplier No.:134500000
Spécifications
À propos de cet article
The protective labeling system provides labels that stand up to harsh laboratory conditions.
- Durable dispenser
- Clear polypropylene tape
- High resistance to chemicals and moisture
- Tape will withstand freezer, autoclave, alkalis, and organic solvents
Durable tape dispenser is molded of strong ABS plastic with a steel serrated cutting blade.
Test results prove that it remains securely affixed with no discoloration after being autoclaved for 40 minutes at 125 °C (258 °F), as well as after one week in liquid nitrogen at –196 °C (–321 °F). All tape sizes are 1mil (0.001") thick with 1mil (0.001") of acrylic adhesive.
White paper labels accept any kind of marker and are perfect for use under the clear tape to label your labware, bottles, or samples. The tape will protect them from the freezer, autoclave, acids, alkalis, and organic solvents.
A tape roll is held in place by an aluminum roller and spring-loaded cover.