Acro® 37 TF Vent Devices, Cytiva (Formerly Pall Lab)
Fournisseur: Cytiva
Certificats
À propos de cet article
Disposable, multipurpose filters for small volume venting, solvent, and gas filtration.
- Disposable
- Multipurpose
- Polypropylene housing
PTFE membrane and polypropylene housing have broad chemical compatibility. Membrane has 0.2 µm pore size and 37 mm diameter with effective filtration area of 7.5 cm². Hose barb connectors are 6.4 to 9.5 mm (¼ to ⅜"). Maximum operating temperature: 100 °C (212 °F). Maximum operating pressure: 4.1 bar (60 psi), bidirectional. Nonsterile.
Frequently Bought Together