Chart Paper, GRAPHIC CONTROLS CO.
Supplier: Graphic Controls
Jump to:
À propos de cet article
This selection of recording chart paper can be used with most popular brand recorders, including those made by both foreign and domestic instrument manufacturers.
10 Options Available Below
Product Details & Documents
This selection of recording chart paper can be used with most popular brand recorders, including those made by both foreign and domestic instrument manufacturers.
Product Family Options
Informations sur le produit
- DésignationPour l'utilisation avecAvailabilityPrice
- Temperature recording chart paper, circular, 7 days, 152 mmUnited Electric Temperature Recorder 650 SeriesDiscontinued-Supplier Number: 00965970Spécifications:
More
Specifications
Cat. No.55476-800 - Supplier Number: 30588969Spécifications:
More
Specifications
Cat. No.CA30588969 - Chart paper, linear, 280 mm×100 ft.Linear recorder 261, 285Log in pour la disponibilitéEach$148.98Supplier Number: 00894634Spécifications:
More
Specifications
Cat. No.CA55476-413 - Chart paper, 237 mm×167 ft.Spectra Physics recorder SP-4270, SP-4290Discontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.CA55476-427 - Supplier Number: 10066240Spécifications:
More
Specifications
Cat. No.CA55476-434 - Supplier Number: 30605532Spécifications:
More
Specifications
Cat. No.CA55476-439 - Supplier Number: 01027341Spécifications:
More
Specifications
Cat. No.CA55476-812 - Supplier Number: 10878487Spécifications:
More
Specifications
Cat. No.CA55476-814 - Chart paper, 64 mm×184 ft.Agilent Technologies recorders 3380A/S, 1080, 5580, 5840A, 5830A, 3885ALog in pour la disponibilitéEach$131.14Supplier Number: 10645035Spécifications:
More
Specifications
Cat. No.CA55476-816 - Chart paper, 208 mm×167 ft.Shimadzu recorder C-R3ARoll of 1$292.56Case of 10$2,925.60Supplier Number: 01062389Spécifications:
More
Specifications
Cat. No.CA55476-425 - Punched chart paper, 208 mm×168 ft.Shimadzu recorder C-R5AEach$405.75Case of 10$4,057.50Supplier Number: 20070365Spécifications:
More
Specifications
Cat. No.CA55476-820 - Temperature recording chart paper, circular, 7 days, 152 mmUnited Electric Temperature Recorder 650 SeriesLog in pour la disponibilitéPack of 50$169.55Supplier Number: 00965970Spécifications:
More
Specifications
Cat. No.CA55476-800