Robotic Tips for Tecan Systems
50 µl, +, Clear, Tecan Systems, 96 per pack, 4800 per case
77507-646
: 332007
NEST robotic tips can be used for research and development and other commonly used applications with needs of high-throughput pipetting in genomics, proteomics, cytomics, immunoassays, metabolomics, biopharmaceuticals.
- Adopt medical grade polypropylene material, which is manufactured by precision mold, and the material batch is stable
- DNase-and RNase-free, no heat source contamination
- Electron beam sterilization
- Manufacturing facility equipped with class 100000 clean-room, products packing under sterile condition
- Unique process technology ensures that the inner surface of the tip is smooth and reduces liquid residue
- Carry out structural drawing and measurement according to the original adapter to ensure that the product has good air tightness and adaptability
- Online testing equipment is used to control the size and concentricity of products in the production line
- Volume:50 µl
- Tip Color:Clear
- Packed:96 per pack, 4800 per case
- Sterile:+
- For Use With:Tecan Systems