Defender™ 3000 Industrial Hybrid Bench Scales
: Ohaus
The OHAUS Defender™ 3000 hybrid - i-D33 bench scales are ideal for general weighing, simple counting and checkweighing applications in production, packaging, warehouse, inventory, shipping and receiving areas.
- Simple, yet rugged, stainless steel design
- Stainless steel indicator with tactile keys
- Multicolor backlit LCD display
- Built in rechargeable battery operation
Results are clearly shown using the large bright backlit LCD display. Display changes color (yellow/green/red) in Checkweighing mode to alert when within the target weight range.
Frequently Bought Together