Pipette Tips
À propos de cet article
NEST 5 and 10 ml pipette tips are precision-molded from high purity, DNase RNase and pyrogen free, virgin polypropylene.
- Autoclavable at 121 ℃
- Meet a wider range of volumetric pipetting needs for more precise pipetting
They are available in 5 and 10 ml, filtered or non-filtered. Choose from compatible pipettors either for Eppendorf, Sartorius, Gilson, or for Thermo pipettors.
They allow pipetting from narrow volume measuring equipment. Clean, hydrophobic surfaces reduce sample retention, and increase accuracy and reproducibility.
Certifications: ISO Class 08.
7 Options Available Below
Product Details & Documents
NEST 5 and 10 ml pipette tips are precision-molded from high purity, DNase RNase and pyrogen free, virgin polypropylene.
- Autoclavable at 121 ℃
- Meet a wider range of volumetric pipetting needs for more precise pipetting
They are available in 5 and 10 ml, filtered or non-filtered. Choose from compatible pipettors either for Eppendorf, Sartorius, Gilson, or for Thermo pipettors.
They allow pipetting from narrow volume measuring equipment. Clean, hydrophobic surfaces reduce sample retention, and increase accuracy and reproducibility.
Certifications: ISO Class 08.
Product Family Options
5 ml tips
Informations sur le produit
- Tip StyleVolumeSterileFilterTip ColorFor Use WithPackedAvailabilityPrice
- Graduated5 ml-ClearFor Thermo100/pack, 1000/caseCase of 1,000$281.29Each$281.29Supplier Number: 307021
- Graduated5 ml++ClearFor Eppendorf, Sartorius, Watson, Gilson240/pack, 1000/caseCase of 720$620.13Each$620.13Supplier Number: 307013
- Graduated5 ml+ClearFor Thermo400/pack, 720/caseCase of 1,200$647.58Each$647.58Supplier Number: 307022
- Graduated5 ml++ClearFor Thermo400/pack, 1200/caseCase of 1,200$894.35Each$894.35Supplier Number: 307023
10 ml tips
Informations sur le produit
- Tip StyleVolumeSterileFilterTip ColorFor Use WithPackedAvailabilityPrice
- Graduated10 ml++ClearFor Eppendof, Sartorius, Gilson240/pack, 720/caseCase of 720$764.14Each$764.14Supplier Number: 307113
- Graduated10 ml+ClearFor Thermo240/pack, 720/caseCase of 720$565.35Each$565.35Supplier Number: 307122
- Graduated10 ml++ClearFor Thermo240/pack, 720/caseCase of 720$757.25Each$757.25Supplier Number: 307123