Defender™ 6000 Industrial Bench Scales
Supplier: Ohaus
Certificates
About this item
Bench scales for standard industrial applications. Defender 6000 indicator constructed of polycarbonate stands up to harsh cleaning in processing and packaging environments with IP68 immersion/IP69K washdown protection.
- Large, bright backlit LCD clearly displays results for fast processing
- The color bar can be used for quick checking of package weights for fast go/no-go alert
- Durably constructed with a 304 stainless steel platform, frame, IP67 loadcell
The Defender 6000 series Hybrid Washdown bench scale is ideal for a multitude of applications, including production and packaging. Defender 6000 handles the most demanding applications with a polycarbonate housing equipped with a 3-color check light bar, ultra-long battery life and EasyConnectTM. The Defender 6000 hybrid is designed to simplify demanding industrial and commercial applications.
Certifications: Defender 6000 hybrid meets NTEP/Measurement Canada approvals.
Frequently Bought Together