Explorer™ Precision Balances
À propos de cet article
The Explorer series is the flagship precision balance range from OHAUS. Equipped with a weighing cell that's precision-machined from a solid metal block, Explorer balances are extremely accurate and durable, and are ideal for laboratory and industrial environments.
- Four-level user management with password protection, and un-editable system log plus a high level of configurability make Explorer balances ideal for regulated applications
- Detachable terminal with a large color touchscreen and programmable IR sensors
- Frameless draftshield make Explorer precision balances easy and convenient to use
The OHAUS Explorer Series of precision balances combine modern features and design elements to offer unmatched functionality in a line of high-performance balances unlike any other on the market. These smart and intuitive balances simplify even the most complex laboratory measurements.
12 Options Available Below
Product Details & Documents
The Explorer series is the flagship precision balance range from OHAUS. Equipped with a weighing cell that's precision-machined from a solid metal block, Explorer balances are extremely accurate and durable, and are ideal for laboratory and industrial environments.
- Four-level user management with password protection, and un-editable system log plus a high level of configurability make Explorer balances ideal for regulated applications
- Detachable terminal with a large color touchscreen and programmable IR sensors
- Frameless draftshield make Explorer precision balances easy and convenient to use
The OHAUS Explorer Series of precision balances combine modern features and design elements to offer unmatched functionality in a line of high-performance balances unlike any other on the market. These smart and intuitive balances simplify even the most complex laboratory measurements.
Product Family Options
Informations sur le produit
- ModèleWeighing CapacityPlateau de pesée/taille de la plateformeReadabilityAvailabilityPrice
- Supplier Number: 30553843
- Supplier Number: 30553844
- Supplier Number: 30553845
- Supplier Number: 30553846
- Supplier Number: 30553847
- Supplier Number: 30553888
- Supplier Number: 30553889
- Supplier Number: 30553890
- Supplier Number: 30553891
- Supplier Number: 30553892
- Supplier Number: 30553893
- Supplier Number: 30553894