Precision Balances, PR
Precision Balances, PR
 :   Ohaus
Precision Balances, PR
 :  Ohaus
Precision Balances, PR
Precision Balances, PR
Designed for routine weighing applications. The PR is equipped with 3 essential weighing modes, RS-232 connectivity for data transfer and storage and internal calibration (in select models), making it ideal for routine weighing applications.
  • Easy-to-read, bright backlit display and a simple user interface
  • Incredibly easy to operate, with almost no training required
  • The small footprint saves desktop space while providing a large weighing surface
  • Durable construction and features a stainless steel pan to withstand day-to-day use in the workplace
Offering accuracy and repeatability in essential weighing applications in laboratory, industrial and education settings, PR Precision Balances deliver competitive performance at an economical price. Featuring RS232 connectivity for easy communication, and a backlit display and a simple interface for uncomplicated operation.
Product Family Options
  • Modèle
    Weighing Capacity
    Plateau de pesée/taille de la plateforme
    Readability
  • PR523/E
    520 g
    Ø 120 mm (4.7")
    0.001 g
    Each
    Each - $2,460.06
    $2,460.06
    Each
       $2,460.06
     : 30430063
  • PR2202/E
    2200 g
    Ø 180 mm (7.1")
    0.01 g
    Each
    Each - $2,167.76
    $2,167.76
    Each
       $2,167.76
     : 30430064
  • PR4202/E
    4200 g
    Ø 180 mm (7.1")
    0.01 g
    Each
    Each - $2,591.20
    $2,591.20
    Each
       $2,591.20
     : 30430065
  • PR4201/E
    4200 g
    Ø 180 mm (7.1")
    0.1 g
    Each
    Each - $1,956.04
    $1,956.04
    Each
       $1,956.04
     : 30430066
  • PR223/E
    220 g
    Ø 120 mm (4.7")
    0.001 g
    Each
    Each - $2,061.90
    $2,061.90
    Each
       $2,061.90
     : 30529239
  • PR1602/E
    1600 g
    Ø 180 mm (7.1")
    0.01 g
    Each
    Each - $1,902.32
    $1,902.32
    Each
       $1,902.32
     : 30529067
  • PR2201/E
    2200 g
    Ø 180 mm (7.1")
    0.1 g
    Each
    Each - $1,658.93
    $1,658.93
    Each
       $1,658.93
     : 30529238