COMPASS™ CR Portable Precision Balances
Supplier: Ohaus
Jump to:
À propos de cet article
Quality portable electronic scales suitable for everyday weighing.
- Intuitive two-button design makes operation easy
- Slim, space-saving design faciliates stackable storage
- Enlarged weighing platform can adequately handle large samples and a variety of objects
The Compass CR series provides basic weighing functionality and competitive performance at an economical price. Features such as a slim, stackable design, and an enlarged weighing platform, make the CR series perfect for a variety of portable weighing applications.
4 Options Available Below
Product Details & Documents
Quality portable electronic scales suitable for everyday weighing.
- Intuitive two-button design makes operation easy
- Slim, space-saving design faciliates stackable storage
- Enlarged weighing platform can adequately handle large samples and a variety of objects
The Compass CR series provides basic weighing functionality and competitive performance at an economical price. Features such as a slim, stackable design, and an enlarged weighing platform, make the CR series perfect for a variety of portable weighing applications.
Product Family Options
Informations sur le produit
- ModèleWeighing CapacityPlateau de pesée/taille de la plateformeReadabilityStabilization TimeAvailabilityPrice
- Supplier Number: 30428204
- Supplier Number: 30428205
- Supplier Number: 30428206
- Supplier Number: 30428207