Universal Pipette Tips
À propos de cet article
Nest Scientific Universal Fit Pipette Tips are made from premium-grade 100% polypropylene and engineered for the most demanding molecular biology applications.
Universal fit: Suitable for use with most pipettors
- Withstands organic solvents
- Less residue
- Made from low retention plastic
Sterilized by gamma irradiation (only valid for some variants), they are certified RNase-, DNase-, human DNA-, and pyrogen-free.
Packaging: Available in different packaging options: Bulk, racked, stack racked or in refills.
15 Options Available Below
Product Details & Documents
Nest Scientific Universal Fit Pipette Tips are made from premium-grade 100% polypropylene and engineered for the most demanding molecular biology applications.
Universal fit: Suitable for use with most pipettors
- Withstands organic solvents
- Less residue
- Made from low retention plastic
Sterilized by gamma irradiation (only valid for some variants), they are certified RNase-, DNase-, human DNA-, and pyrogen-free.
Packaging: Available in different packaging options: Bulk, racked, stack racked or in refills.
Product Family Options
Bulk, non sterile
Informations sur le produit
- Tip StyleVolumeSterileTip ColorPackedAvailabilityPrice
- Graduated, extra long10 µl-Clear1000/pack, 10000/caseCase of 10,000$294.51Each$294.51Supplier Number: 304001
- Graduated1250 µl (1,25 ml)-Clear1000/pack, 10000/caseCase of 10,000$542.41Each$542.41Supplier Number: 306001
Racked, sterile
Informations sur le produit
- Tip StyleVolumeSterileTip ColorPackedAvailabilityPrice
- Graduated10 µl+Clear96/box, 4800/caseCase of 4,800$389.63Each$389.63Supplier Number: 301012
- Graduated, extra long10 µl+Clear96/box, 4800/caseCase of 4,800$389.74Each$389.74Supplier Number: 304012
- Graduated200 µl+Clear96/box, 4800/caseCase of 4,800$425.27Each$425.27Supplier Number: 302012
- Graduated300 µl+Clear96/box, 4800/caseCase of 4,800$442.03Each$442.03Supplier Number: 305012
- Graduated1250 µl (1,25 ml)+Clear96/box, 4800/caseCase of 4,800$572.00Each$572.00Supplier Number: 306012
Refills, non sterile
Informations sur le produit
- Tip StyleVolumeSterileTip ColorPackedAvailabilityPrice
- Graduated, extra long10 µl-Clear96/rack, 960/pack, 9600 caseCase of 9,600$528.12Each$528.12Supplier Number: 304023
- Graduated300 µl-Clear96/rack, 960/pack, 9600 caseCase of 9,600$546.41Each$546.41Supplier Number: 305023
- Graduated1250 µl (1,25 ml)-Clear96/rack, 960/pack, 9600 caseCase of 4,800$312.73Each$312.73Supplier Number: 306023
Double stack racked, sterile
Informations sur le produit
- Tip StyleVolumeSterileTip ColorPackedAvailabilityPrice
- Graduated10 µl+Clear96/wafer, 960/pack, 9600 caseCase of 9,600$535.76Each$535.76Supplier Number: 301052
- Graduated, extra long10 µl+Clear96/wafer, 960/pack, 9600 caseCase of 9,600$535.66Each$535.66Supplier Number: 304052
- Graduated200 µl+Clear96/wafer, 960/pack, 9600 caseCase of 9,600$575.08Each$575.08Supplier Number: 302052
Bagged, non sterile
Informations sur le produit
- Tip StyleVolumeSterileTip ColorPackedAvailabilityPrice
- Graduated200 µl-Jaune1000/pack, 20000/caseCase of 20,000$515.79Each$515.79Supplier Number: 302106
- Graduated1000 µl-Bleu1000/pack, 5000/caseCase of 50,000$290.90Each$290.90Supplier Number: 303206