CELLCOAT® SCREENSTAR MultiCoat Test Plates
Test plates designed to help users find the optimal cell culture surface for assays.
- Offers 11 different peptide and protein coatings on a single microplate to test various biological surfaces simultaneously
- Equipped with a cycloolefin bottom that has glass-like optical properties and planarity for optimal image quality
- Available in 96-well and 384-well formats to accommodate different assay needs and throughput requirements
- Improves cell attachment, spreading, distribution, and growth, and can modulate differentiation and protein expression
- Allows for quick and straightforward testing of multiple biological coatings to find the most suitable in-vivo mimicking environment for specific cells
The multiple biomolecular coatings enhance binding capacity and specificity, making multicoat plates ideal for diverse high-throughput applications. Both 96- and 384-well formats are compatible with automated systems, ensuring seamless integration into laboratory workflows.
: Ask about black CELLCOAT® SCREENSTAR plates with cycloolefin available in 96, 384, and 1536-well formats.
: Packaged 2 pieces per bag (plate and lid).
Test plates designed to help users find the optimal cell culture surface for assays.
- Offers 11 different peptide and protein coatings on a single microplate to test various biological surfaces simultaneously
- Equipped with a cycloolefin bottom that has glass-like optical properties and planarity for optimal image quality
- Available in 96-well and 384-well formats to accommodate different assay needs and throughput requirements
- Improves cell attachment, spreading, distribution, and growth, and can modulate differentiation and protein expression
- Allows for quick and straightforward testing of multiple biological coatings to find the most suitable in-vivo mimicking environment for specific cells
The multiple biomolecular coatings enhance binding capacity and specificity, making multicoat plates ideal for diverse high-throughput applications. Both 96- and 384-well formats are compatible with automated systems, ensuring seamless integration into laboratory workflows.
: Ask about black CELLCOAT® SCREENSTAR plates with cycloolefin available in 96, 384, and 1536-well formats.
: Packaged 2 pieces per bag (plate and lid).
Product Family Options
- Nb de puitsWorking VolumeRevêtementWell Shape
- 9625 - 440 μl11 peptide and protein coatings (single and double coatings)F-bottom/chimney wellCase of 2$1,152.43: 655866-MCP
- 38410 - 110 μl11 peptide and protein coatings (single and double coatings)F-bottom wellCase of 2$1,256.55: 781866-MCP