Laboratory Software, Labworldsoft 6 | Avantor
Laboratory Software, Labworldsoft 6
Labworldsoft® 6 Visc, for viscosity measurements for thermostats and pH meters of all manufacturers
Catalog # 76499-030
Supplier:   IKA Works
Fournisseur #:  0020101872
Laboratory Software, Labworldsoft 6
Catalog # 76499-030
Supplier:  IKA Works
Laboratory Software, Labworldsoft 6
Laboratory Software, Labworldsoft 6
Labworldsoft® 6 Visc, for viscosity measurements for thermostats and pH meters of all manufacturers
Price
$2,734.69
Log in pour la disponibilité
Each
Product Details & Documents
Labworldsoft 6 is a laboratory software to control lab devices, automate processes, and to document your measurements and results. The software provides access to a huge library of different laboratory devices.
  • Easy recording of various physical parameters
  • Function modules to create time triggered or event triggered configurations
  • Communication through RS232, USB, Bluetooth or network
  • Supports formats .csv, .xslx and ASCII
  • Compatibility with Windows 7, 8 and 9
Labworldsoft does not integrate only IKA laboratory devices, but supports several other device from different manufacturers as well. You stay flexible in the choice of your lab devices you want to use and profit from the whole device range of the laboratory world.
 : Includes one year of support with new versions of Labworldsoft 6.
Spécifications
  • Désignation:
    Labworldsoft® 6 Visc, for viscosity measurements for thermostats and pH meters of all manufacturers