Digital Force Meter | Avantor
Digital Force Meter
Supplier:   GSC International
Digital Force Meter
Supplier:  GSC International
Certificates
Digital Force Meter
Digital Force Meter
2 Options Available Below
Product Details & Documents
Ideal for addition and resolution of forces, Newton's laws, and statics experiments.
  • Wide measureable range and decimal resolution makes this instrument a versatile alternative to traditional spring scales
With a wide range of 2, 5, 10 and 20 Newtons, this force meter replaces four types of spring balances. With the hold function, you can retain the measured value for added convenience. With the one-touch zero-calibration function, you can measure in any direction, 360 degrees, for complete accuracy. Compact and convenient, this digital force meter measures 157 mm in length, 45 mm in width, and 24 mm in height, making it easy to use and store.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Digital force meter, 0.1 - 20 N, pack of 10
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $1,185.00
    $1,185.00
    Pack of 10
       $1,185.00
    Supplier Number: N-00001-10
  • Digital force meter, 0.1 - 20 N, single
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $123.25
    $123.25
    Each
       $123.25
    Supplier Number: N-00001